GeneJET PCR Purification Kit, 250 preps - FAQs
GeneJET PCR Purification Kit, 250 preps - FAQs
View additional product information for GeneJET PCR Purification Kit - FAQs (K0702, K0701)
7 product FAQs found
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?
We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.
Can I use a vacuum manifold with GeneJET kits?
The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.
Can I use GeneJET PCR Purification Kit for DNA gel extraction?
GeneJET PCR Purification Kit is designed for PCR reaction clean-up. The kit can be used for gel extraction by following the protocol of GeneJET Gel Extraction Kit, but DNA yield may be compromised. For best performance of DNA gel extraction, we recommend to use GeneJET Gel Extraction Kit. For a 3-in-1 kit for purification of PCR products from enzymatic reaction mixtures and agarose gels, we recommend GeneJET Gel Extraction and DNA Cleanup Micro Kit (Cat. Nos. K0831/2).
Can I use GeneJET PCR Purification Kit for enzymatic clean-up?
GeneJET PCR Purification Kit can be used for enzymatic clean-up. Please follow the PCR reaction clean-up protocol.
At what temperature should I perform centrifugation steps in the GeneJET kits?
We recommend performing centrifugation steps at room temperature when the temperature is not specified.
Are GeneJET spin columns or MagJET magnetic beads available separately?
No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.
Do you offer the binding buffer included in the GeneJET PCR Purification Kit as a stand-alone product?
Yes, we offer Binding Buffer for GeneJET Gel Extraction Kit as a stand-alone product: Cat. No. R1331.