Maxima Hot Start PCR Master Mix (2X) - FAQs
Maxima Hot Start PCR Master Mix (2X) - FAQs
View additional product information for Maxima Hot Start PCR Master Mix (2X) - FAQs (K1052)
1 product FAQs found
Can I do PCR directly from bacterial colonies with Thermo Scientific Taq Master Mixes?
Dream Taq PCR Master Mix and Taq PCR Master Mix can be used for direct PCR from bacterial colonies. Simply scrape a tiny amount of a single colony and transfer to a PCR-tube, prefilled with the master mix. Always use at least 3 min at 95 degrees C for initial denaturation.