DreamTaq Green PCR Master Mix (2X), 200 Reactions - FAQs
DreamTaq Green PCR Master Mix (2X), 200 Reactions - FAQs
View additional product information for DreamTaq PCR Master Mixes (2X) - FAQs (K1072, K1071, K1082, K1081)
1 product FAQs found
What is the error rate of DreamTaq DNA Polymerases?
The error rate of DreamTaq DNA Polymerase is the same as that of Taq DNA Polymerase: 2.2x10^-5 errors per nt per cycle.