Rapid DNA Ligation Kit
Thermo Scientific™

Rapid DNA Ligation Kit

Thermo Scientific Rapid DNA Ligation Kit enables fast sticky-end or blunt-end DNA ligation in only 5 minutes at room temperature.TheRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
K142250 Reactions
K1423150 Reactions
Catalog number K1422
Price (MXN)
-
Quantity:
50 Reactions
Request bulk or custom format
Thermo Scientific Rapid DNA Ligation Kit enables fast sticky-end or blunt-end DNA ligation in only 5 minutes at room temperature.The kit contains T4 DNA ligase and a specially-formulated 5X rapid ligation buffer optimized for fast and efficient DNA ligation. Fast ligation efficiency is equal to that obtained with T4 DNA ligase in a standard 1 hour ligation. The ligation reaction mixture can be used directly for bacterial transformation with the TransformAid Bacterial Transformation Kit or with other conventional transformation procedures.

Highlights

• Fast—ligation of either sticky- or blunt-end DNA in only 5 minutes
• Convenient—reaction mixture can be used directly for bacterial transformation

Applications

• Routine cloning experiments
• Blunt-end cloning
• Library construction
• TA cloning

Includes

• T4 DNA Ligase
• 5X Rapid Ligation Buffer
• Water, nuclease-free
• Detailed Protocol

Note

Prior to electroporation, it is necessary to inactivate T4 DNA ligase by chloroform extraction or spin column purification, e.g. with Thermo Scientific GeneJET PCR Purification Kit (K0701).

Related Products
Rapid DNA Ligation Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Buffer5X Rapid Ligation Buffer
EnzymeDNA Ligase
Product TypeRapid DNA Ligation Kit
Quantity50 Reactions
Unit SizeEach

Frequently asked questions (FAQs)

What is the difference between Rapid DNA Ligation Kit and Thermo Scientific T4 DNA ligase with a standard buffer?

The speed of ligation is the main difference between the two. The Thermo Scientific Rapid DNA Ligation Kit enables fast sticky-end or blunt-end DNA ligation in only 5 minutes at room temperature. The kit contains T4 DNA ligase and a specially-formulated 5X rapid ligation buffer optimized for fast and efficient DNA ligation. Fast ligation efficiency is equal to that obtained with T4 DNA ligase in a standard 1 hour ligation.