PureLink™ miRNA Isolation Kit - FAQs
PureLink™ miRNA Isolation Kit - FAQs
View additional product information for PureLink™ miRNA Isolation Kit - FAQs (K157001)
1 product FAQs found
Have you tested the PureLink RNA Mini Kit (Cat. No. 12183018A) or PureLink miRNA Isolation Kit (Cat. No. K157001) for extraction of RNA with the TaqMan Advanced miRNA Assays?
These kits have not been tested. However, these kits should work with the TaqMan Advanced miRNA Assays. We recommend checking the quality and quantity of small RNAs post-isolation by NanoDrop and Bioanalyzer.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.