mRNA Catcher™ PLUS Purification Kit, 96 preps - FAQs
mRNA Catcher™ PLUS Purification Kit, 96 preps - FAQs
View additional product information for mRNA Catcher™ PLUS Purification Kit - FAQs (K157004, K157003, K157002)
3 product FAQs found
What types of samples can be used to isolate mRNA with the mRNA Catcher PLUS?
The mRNA Catcher PLUS kit is designed to isolate mRNA from cells, animal tissues, whole blood, and total RNA.
How many cells should I use for RNA isolation with the mRNA CatcherPLUS Kit?
We recommend using a minimum of 100 cells per well for mRNA isolation with the mRNA CatcherPLUS Kit.
Which PCR machines is the plate in mRNA CatcherPLUS Kit compatible with?
The mRNA CatcherPLUS Kit plate should fit in any standard 96-well PCR machine including MJ and ABI thermocyclers.