PureLink™ PCR Purification Kit, 250 preps - FAQs
PureLink™ PCR Purification Kit, 250 preps - FAQs
View additional product information for PureLink™ PCR Purification Kit - FAQs (K310002, K310001)
3 product FAQs found
Can I use the PureLink PCR Purification Kit to clean up restriction digested plasmid DNA?
Yes, you can use our kit to clean up restriction digested plasmid DNA.
Can I use the PureLink PCR Purification Kit to do a gel purification?
The buffers and procedure are different for the PureLink PCR Purification Kit and PureLink Gel Purification Kit. Therefore, you can swap the column, but the right buffer and protocol must be used.
What are the minimum and maximum size ranges for DNA fragment cleanup using the PureLink PCR Clean-Up Kit?
The size range is between 100 bp and 12 kb. Our kit comes with two buffers, Binding Buffer (B2) and Binding Buffer HC (B3). The Binding Buffer HC (B3) eliminates primer-dimers and short failed PCR products that are smaller than 300 bp.