His-Patch ThioFusion™ Expression System - FAQs
His-Patch ThioFusion™ Expression System - FAQs
View additional product information for His-Patch ThioFusion™ Expression System - FAQs (K36001)
1 product FAQs found
During protein purification, is it typical for material to remain insoluble in 6M guanidinium lysis buffer?
It is not unusual for insoluble material to remain in the lysate. The expressed protein should be in the supernatant. Extremely insoluble proteins may be solubilized by including 0.1% sarkosyl (n-lauryl sarcosinate) in the lysis buffer.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.