Product Image
Invitrogen™

pLenti6/V5 Directional TOPO™ Cloning Kit

The pLenti6⁄V5 Directional TOPO™ Cloning Kit contains the TOPO™-adapted ViraPower™ lentiviral expression vector, pLenti6⁄V5-D-TOPO™ for quick PCR-based cloning and high-levelRead more
Have Questions?
Catalog NumberQuantity
K49551020 Reactions
Catalog number K495510
Price (USD)
3,090.00
Each
Quantity:
20 Reactions
Price (USD)
3,090.00
Each
The pLenti6⁄V5 Directional TOPO™ Cloning Kit contains the TOPO™-adapted ViraPower™ lentiviral expression vector, pLenti6⁄V5-D-TOPO™ for quick PCR-based cloning and high-level expression of a target gene in dividing and non-dividing mammalian cells. The vector has the CMV promoter for driving high-level, constitutive expression of the target gene and the blasticidin selection marker for stable selection in mammalian cells.

Advantages
• High efficiency and rapid cloning
• Constitutive gene expression in dividing and non-dividing mammalian cells in vitro or in vivo
• Produces replication-incompetent virus for enhanced biosafety of the system

Key Features
• Directional TOPO™ Cloning site for rapid and efficient directional cloning of blunt-end PCR products
• Rouse Sarcoma Virus (RSV) enhancer⁄promoter for Tat-independent production of viral mRNA in the producer cell line
• Modified HIV-1 5’ and 3’ Long Terminal Repeats (LTR) for viral packaging and reverse transcription
• HIV-1 psi (ψ) packaging sequence for viral packaging
• HIV Rev response element (RRE) for Rev-dependent nuclear export of unspliced viral mRNA
• (CMV) immediate early promoter for high-level constitutive expression of the gene of interest in mammalian cells
• C-terminal V5 epitope for detection of the recombinant protein
• Blasticidin (bsd) resistance gene for selection in E. coli and mammalian cells
• Ampicillin resistance gene for selection in E. coli
• pUC origin for high-copy replication and maintenance of the plasmid in E. coli

Kit includes
pLenti6⁄V5-D- TOPO™ Reagents
One Shot™ Stbl3™ Chemically Competent E. coli

For research use only. Not intended for any therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cloning MethodDirectional TOPO™
Constitutive or Inducible SystemConstitutive
Delivery TypeLentiviral
For Use With (Application)Viral Expression
No. of Reactions20 reactions
Product LineTOPO, ViraPower
Product TypeTOPO Cloning Kit
PromoterCMV
Protein TagV5 Epitope Tag
Quantity20 Reactions
Selection Agent (Eukaryotic)Blasticidin
VectorpLenti, Directional TOPO Vectors
Unit SizeEach
Contents & Storage
pLenti⁄V5-D-TOPO™ reagents (Box 1) Store at -20°C
• pLenti6⁄V5-D-TOPO™ Reagents (15-20 ng⁄μl linearized plasmid DNA in: 50% glycerol, 50 mM Tris-HCl, pH 7.4 (at 25°C),1 mM EDTA, 2 mM DTT, 0.1% Triton X-100, 100 μg⁄ml BSA, 30 μM bromophenol blue)
• dNTP Mix (12.5 mM each dATP, dCTP, dGTP, dTTP in water, pH8 at a total volume of 10 μl)
• Salt Solution (1.2 M NaCl and 0.06 M MgCl2, volume of 50 μl)
• Water (1 ml)
• CMV Forward Sequencing Primer (0.1 μg⁄μl in TE Buffer, pH 8, pH8 at a total volume of 20 μl)
• Control PCR Primers (0.1 μg⁄μl each in TE Buffer, pH 8, pH8 at a total volume of 10 μl)
• Control PCR Template (0.1 μg⁄μl in TE Buffer, pH 8, pH8 at a total volume of 10 μl)
• pLenti6⁄V5-GW⁄lacZ (lyophilized in TE Buffer, pH 8, 10 μg)

One Shot™ Stbl3™ Chemically Competent E. coli (Box 2) Store at -80°C
• S.O.C. Medium (2% Trypton, 0.5% Yeast Extract, 10 mM NaCl, 2.5 mM KCl, 10 mM MgCl2, 10 mM MgSO4, 20 mM glucose. 6ml) May be stored at +4°C or room temperature
• Stbl3™ Cells (21 x 50 μl)
• pUC19 Control DNA (10 pg⁄μl in 5 mM Tris-HCl, 0.5 mM EDTA, pH 8. 50 μl)

Frequently asked questions (FAQs)

I performed stable selection but my antibiotic-resistant clones do not express my gene of interest. What could have gone wrong?

Here are possible causes and solutions:

Detection method may not be appropriate or sensitive enough:
- We recommend optimizing the detection protocol or finding more sensitive methods. If the protein is being detected by Coomassie/silver staining, we recommend doing a western blot for increased sensitivity. The presence of endogenous proteins in the lysate may obscure the protein of interest in a Coomassie/silver stain. If available, we recommend using a positive control for the western blot.
- Insufficient number of clones screened: Screen at least 20 clones.
- Inappropriate antibiotic concentration used for stable selection: Make sure the antibiotic kill curve was performed correctly. Since the potency of a given antibiotic depends upon cell type, serum, medium, and culture technique, the dose must be determined each time a stable selection is performed. Even the stable cell lines we offer may be more or less sensitive to the dose we recommend if the medium or serum is significantly different.
- Expression of gene product (even low level) may not be compatible with growth of the cell line: Use an inducible expression system.
- Negative clones may result from preferential linearization at a vector site critical for expression of the gene of interest: Linearize the vector at a site that is not critical for expression, such as within the bacterial resistance marker.

I used a mammalian expression vector but do not get any expression of my protein. Can you help me troubleshoot?

Here are possible causes and solutions:

- Try the control expression that is included in the kit
Possible detection problem:

- Detection of expressed protein may not be possible in a transient transfection, since the transfection efficiency may be too low for detection by methods that assess the entire transfected population. We recommend optimizing the transfection efficiency, doing stable selection, or using methods that permit examination of individual cells. You can also increase the level of expression by changing the promoter or cell type.
- Expression within the cell may be too low for the chosen detection method. We recommend optimizing the detection protocol or finding more sensitive methods. If the protein is being detected by Coomassie/silver staining, we recommend doing a western blot for increased sensitivity. The presence of endogenous proteins in the lysate may obscure the protein of interest in a Coomassie/silver stain. If available, we recommend using a positive control for the western blot. Protein might be degraded or truncated: Check on a Northern. Possible time-course issue: Since the expression of a protein over time will depend upon the nature of the protein, we always recommend doing a time course for expression. A pilot time-course assay will help to determine the optimal window for expression. Possible cloning issues: Verify clones by restriction digestion and/or sequencing.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

I am using a mammalian expression vector that has the neomycin resistance gene. Can I use neomycin for stable selection in mammalian cells?

No; neomycin is toxic to mammalian cells. We recommend using Geneticin (a.k.a. G418 Sulfate), as it is a less toxic and very effective alternative for selection in mammalian cells.

Is it okay if my construct has an ATG that is upstream of the ATG in my gene of interest? Will it interfere with translation of my gene?

Translation initiation will occur at the first ATG encountered by the ribosome, although in the absence of a Kozak sequence, initiation will be relatively weak. Any insert downstream would express a fusion protein if it is in frame with this initial ATG, but levels of expressed protein are predicted to be low if there is a non-Kozak consensus sequence. If the vector contains a non-Kozak consensus ATG, we recommend that you clone your gene upstream of that ATG and include a Kozak sequence for optimal expression.

Do you offer a GFP-expressing mammalian expression vector that I can use as a control to monitor my transfection and expression?

We offer pJTI R4 Exp CMV EmGFP pA Vector, Cat. No. A14146, which you can use to monitor your transfection and expression.

Citations & References (2)

Citations & References
Abstract
RIP140-targeted repression of gene expression in adipocytes.
Authors:Christian M, Kiskinis E, Debevec D, Leonardsson G, White R, Parker MG,
Journal:Mol Cell Biol
PubMed ID:16227589
'Ligand-dependent repression of nuclear receptor activity forms a novel mechanism for regulating gene expression. To investigate the intrinsic role of the corepressor RIP140, we have monitored gene expression profiles in cells that express or lack the RIP140 gene and that can be induced to undergo adipogenesis in vitro. In contrast ... More
Coregulation of vascular tube stabilization by endothelial cell TIMP-2 and pericyte TIMP-3.
Authors:Saunders WB, Bohnsack BL, Faske JB, Anthis NJ, Bayless KJ, Hirschi KK, Davis GE,
Journal:J Cell Biol
PubMed ID:17030988
The endothelial cell (EC)-derived tissue inhibitor of metalloproteinase-2 (TIMP-2) and pericyte-derived TIMP-3 are shown to coregulate human capillary tube stabilization following EC-pericyte interactions through a combined ability to block EC tube morphogenesis and regression in three-dimensional collagen matrices. EC-pericyte interactions strongly induce TIMP-3 expression by pericytes, whereas ECs produce TIMP-2 ... More