Product Image
Invitrogen™

pENTR™ 5'-TOPO™ TA Cloning™ Kit, with One Shot™ TOP10 Chemically Competent E. coli

The pENTR™ 5’-TOPO™ TA Cloning™ Kit combines Invitrogen’s TOPO™ Cloning and MultiSite Gateway™ Technologies to facilitate five-minute, one-step cloning ofRead more
Have Questions?
Catalog NumberQuantity
K5912020 reactions
Catalog number K59120
Price (USD)
752.94
Special offer
Online exclusive
Ends: 27-Jun-2026
846.00
Save 93.06 (11%)
Each
Quantity:
20 reactions
Price (USD)
752.94
Special offer
Online exclusive
Ends: 27-Jun-2026
846.00
Save 93.06 (11%)
Each
The pENTR™ 5’-TOPO™ TA Cloning™ Kit combines Invitrogen’s TOPO™ Cloning and MultiSite Gateway™ Technologies to facilitate five-minute, one-step cloning of a Taq polymerase-amplified PCR product encoding a eukaryotic promoter of interest into a MultiSite Gateway™ entry vector with ≥ 85% efficiency. Once cloned, your eukaryotic promoter may be transferred from the pENTR™ 5'-TOPO™ vector to a suitable MultiSite Gateway™ destination vector in a MultiSite Gateway™ LR recombination reaction with other suitable entry clone(s) to generate an expression construct.

This kit can be used in combination with ViraPower™ HiPerform Promoterless Lentiviral Gateway™ Vector Kit, or MultiSite Gateway™ Three-Fragment Vector Construction Kit.

Advantages
• Choose the best promoter for your experiments and cell line by efficiently expressing your gene of interest from multiple promoter constructs
• When using in conjunction with ViraPower™ HiPerform™ Promoterless Lentiviral vector, you can efficiently transduce and express your gene of interest driven by the promoter of choice in both dividing and non-dividing mammalian cells


Key Features
• Rapid PCR cloning with TOPO™ cloning technology for cloning the 5’ element
• attL4 & attR1 sites for compatibility with MultiSite Gateway™ LR reaction for increased flexibility

Kit Includes
• pENTR™ 5´-TOPO™ TA Cloning™ Kit
• One Shot™ TOP10 Chemically Competent E. coli (C4040-03)

Related SKUs
• ViraPower™ HiPerform™ Promoterless Lentiviral Gateway™ Expression System (A111451)
• ViraPower™ HiPerform™ Promoterless Gateway™ Vector Kit (A11146)
• MultiSite Gateway™ Three-Fragment Vector Construction Kit (12537-023)

For research use only. Not intended for any therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeTA Cloning Kit
Quantity20 reactions
VectorpENTR, TOPO-TA Cloning Vectors
Cloning MethodTOPO-TA
Product LineOne Shot
Unit SizeEach
Contents & Storage
The pENTR™ 5´-TOPO™ TA Cloning™ Kit with One Shot™ TOP10 Chemically Competent E. coli contains two boxes.

The TOPO™ Cloning box contains enough linearized and topoisomerase I-activated pENTR™ 5´-TOPO™ vector for 20 cloning reactions, dNTPs, salt solution, control template and primers and primers for sequencing. Store at -20°C.

The One Shot™ TOP10 box contains transformation reagents including 20 single-use 50 μl aliquots of One Shot™ Competent Cells, S.O.C. medium, and pUC19 supercoiled plasmid control. Store at -80°C.

Frequently asked questions (FAQs)

Can I store my competent E. coli in liquid nitrogen?

We do not recommend storing competent E. coli strains in liquid nitrogen as the extreme temperature can be harmful to the cells. Also, the plastic storage vials are not intended to withstand the extreme temperature and may crack or break.

How should I store my competent E. coli?

We recommend storing our competent E. coli strains at -80°C. Storage at warmer temperatures, even for a brief period of time, will significantly decrease transformation efficiency.

Your Gateway-adapted TOPO vectors are supplied with a control template and control primers. Can I obtain the sequence of the control template?

The sequence of the control template is proprietary.

What is the best molar ratio of PCR product:vector to use for TOPO TA cloning? Is there an equation to calculate the quantity to use?

We suggest starting with a molar ratio of 1:1 (insert:vector), with a range of 0.5:1 to 2:1. The quantity used in a TOPO cloning reaction is typically 5-10 ng of a 2 kb PCR product.

Equation:

length of insert (bp)/length of vector (bp) x ng of vector = ng of insert needed for 1:1 (insert:vector ratio)

What is the best ratio of insert:vector to use for cloning? Is there an equation to calculate this?

The optimal ratio is 1:1 insert to vector. Optimization can be done using a ratio of 0.5-2 molecules of insert for every molecule of the vector.

Equation:

length of insert (bp)/length of vector (bp) x ng of vector = ng of insert needed for 1:1 insert:vector ratio