Ni-NTA Purification System with Antibody, with Anti-His (C-term)-HRP Antibody - FAQs
Ni-NTA Purification System with Antibody, with Anti-His (C-term)-HRP Antibody - FAQs
View additional product information for Ni-NTA Purification System with Antibody, with Anti-His (C-term)-HRP Antibody - FAQs (K95301)
2 product FAQs found
Can ProBond or Ni-NTA beads be used for large-scale preparations?
ProBond and Ni-NTA beads can be used in FPLC columns. However, the beads can only withstand low pressure (~43.5 psi max).
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What should the typical protein recovery be when using the Probond Purification System or Ni-NTA Purification system?
Both systems are qualified by purifying 2 mg of myoglobin protein on a column and performing a Bradford assay. Protein recovery must be 75% or higher.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.