TA Cloning® Kit, with pCR™2.1 Vector and One Shot® TOP10F' Chemically Competent E. coli, no manual - FAQs
TA Cloning® Kit, with pCR™2.1 Vector and One Shot® TOP10F' Chemically Competent E. coli, no manual - FAQs
View additional product information for TA Cloning® Kit, with pCR™2.1 Vector and One Shot® TOP10F' Chemically Competent E. coli, no manual - FAQs (KNM203001)
1 product FAQs found
Can I use TOP10F' competent cells for transformation of my TOPO vector that contains the ccdB gene?
Strains that contain an F plasmid, such as TOP10F', are not recommended for transformation and selection of recombinant clones in any TOPO vector containing the ccdB gene. While the F plasmid does encode the CcdA protein, which acts as an inhibitor of the CcdB gyrase-toxin protein, the half-life of the CcdA protein is shorter than that of the CcdB protein. Overexpression of the CcdB protein causes cell death when its action is not prevented by sufficient CcdA.