(S)-(-)-2-Phényl-1-propanol, 98+ %, Thermo Scientific Chemicals
(S)-(-)-2-Phényl-1-propanol, 98+ %, Thermo Scientific Chemicals
(S)-(-)-2-Phényl-1-propanol, 98+ %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-(-)-2-Phényl-1-propanol, 98+ %, Thermo Scientific Chemicals

CAS: 37778-99-7 | C9H12O | 136.194 g/mol
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
250 mg
1 g
Référence L13988.03
également connu sous le numéro L13988-03
Prix (EUR)
197,00
Each
Quantité:
1 g
Grand volume ou format personnalisé
Prix (EUR)
197,00
Each
Identifiants chimiques
CAS37778-99-7
IUPAC Name2-phenylpropan-1-ol
Molecular FormulaC9H12O
InChI KeyRNDNSYIPLPAXAZ-UHFFFAOYNA-N
SMILESCC(CO)C1=CC=CC=C1
Voir plus
FormLiquid
Appearance (Color)Clear colorless
Optical Rotation-17.3 ± 0.1? (neat)
Refractive Index1.5245-1.5285 @ 20?C
Assay (GC)≥98.0%
For synthesis of optically active products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only