Lectin Helix pomatia agglutinin (HPA) selectively binds to α-N-acetylgalactosamine residues and type A erythrocytes.View complete list of fluorescent dye-conjugated lectinsRead more
Have Questions?
Catalog Number
Quantity
L32454
1 mg
Catalog number L32454
Price (USD)
285.36
Special offer
Online exclusive
Ends: 31-Dec-2026
348.00
Save 62.64
Each
Quantity:
1 mg
Price (USD)
285.36
Special offer
Online exclusive
Ends: 31-Dec-2026
348.00
Save 62.64
Each
Lectin Helix pomatia agglutinin (HPA) selectively binds to α-N-acetylgalactosamine residues and type A erythrocytes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label TypeAlexa Fluor Dyes
Product LineAlexa Fluor
Protein SubtypeLectins
Quantity1 mg
Shipping ConditionRoom Temperature
SourceEdible snail
ConjugateAlexa Fluor 647
FormLyophilized
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Citations & References (11)
Citations & References
Abstract
Biochemical and structural analysis of Helix pomatia agglutinin. A hexameric lectin with a novel fold.
Authors:Sanchez JF, Lescar J, Chazalet V, Audfray A, Gagnon J, Alvarez R, Breton C, Imberty A, Mitchell EP
Journal:J Biol Chem
PubMed ID:16704980
'Helix pomatia agglutinin (HPA) is a N-acetylgalactosamine (GalNAc) binding lectin found in the albumen gland of the roman snail. As a constituent of perivitelline fluid, HPA protects fertilized eggs from bacteria and is part of the innate immunity system of the snail. The peptide sequence deduced from gene cloning demonstrates ... More
Digging deep into Golgi phenotypic diversity with unsupervised machine learning.
Authors:
Journal:Mol Biol Cell
PubMed ID:29021342
The Effect of Hypoxia and Metformin on Fatty Acid Uptake, Storage, and Oxidation in L6 Differentiated Myotubes.
Authors:
Journal:Front Endocrinol (Lausanne)
PubMed ID:30386299
The GalNAc-T Activation (GALA) Pathway: Drivers and markers.
Authors:
Journal:PLoS One
PubMed ID:30889231
RNAi screening reveals a large signaling network controlling the Golgi apparatus in human cells.