Dual LED Blue/White Light Transilluminator - FAQs
Dual LED Blue/White Light Transilluminator - FAQs
View additional product information for Dual LED Blue/White Light Transilluminator - FAQs (LB0100)
8 product FAQs found
Does the Dual LED Blue/White Light Transilluminator (Cat. No. LB0100) come with plug in adapters for different global regions?
Yes! The transilluminator will be shipped with the respective plug in adapters for the respective region.
Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.
For the Dual LED Blue/White Light Transilluminator (Cat. No. LB0100), can I purchase any part of this product as standalone item?
No, no parts are sold separately for this product.
Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.
Can I use the Dual LED Blue/White Light Transilluminator to visualize gels stained with SYBR Safe DNA Gel Stain or SYBR Gold Nucleic Acid Gel Stain?
Yes, the Dual LED Blue/White Light Transilluminator can be used to visualize gels stained with SYBR Safe DNA Gel Stain, SYBR Gold Nucleic Acid Gel Stain, or Ethidium bromide.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.
Can I visualize ethidium bromide-stained gels with the Dual LED Blue/White Light Transilluminator?
Ethidium bromide-stained gels can be visualized with the Dual LED Blue/White Light Transilluminator as ethidium bromide has a smaller secondary excitation peak in the range of 400-570 nm. However, keep in mind that the fluorescence intensity will not be as bright as when excited at the primary excitation peak of 300 nm, that would be possible with UV transilluminators.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
For the Dual LED Blue/White Light Transilluminator, can I disable the automatic power-off function?
The 5 min automatic power-off function cannot be disabled, but the instrument can be turned on again immediately after it turns off.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
For the Dual LED Blue/White Light Transilluminator, can I purchase the white/blue plates and imaging hood as standalone items?
The white/blue plates and imaging hood are not sold separately.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can the automatic power-off be disabled for the Dual LED Blue/White Light Transilluminator?
The 5 min automatic power-off cannot be disabled, but the instrument can be turned back on again right away.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
I have the Dual LED Blue/White Light Transilluminator. Are the white/blue plates and imaging hood sold separately?
White/blue plates and the imaging hood are not sold separately.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.