Novex™ Tris-Glycine SDS Running Buffer (10X)
Invitrogen™

Novex™ Tris-Glycine SDS Running Buffer (10X)

Novex Tris-Glycine SDS Running Buffer (10X) is formulated for separation of proteins in their denatured state on Tris-Glycine gels. Tris-GlycineRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LC267555 L
LC2675500 mL
LC267544 x 1 L
Catalog number LC26755
Price (USD)
141.65
Online exclusive
146.00
Save 4.35
Each
Quantity:
5 L
Price (USD)
141.65
Online exclusive
146.00
Save 4.35
Each
Novex Tris-Glycine SDS Running Buffer (10X) is formulated for separation of proteins in their denatured state on Tris-Glycine gels. Tris-Glycine gels provide reproducible separation of a wide range of proteins into well-resolved bands.

Separate native or denatured proteins
Novex Tris-Glycine gels do not contain SDS and can be used to accurately separate both native and denatured proteins, depending upon the sample and running buffers used. To separate denatured proteins on Novex Tris-Glycine gels, use Novex Tris-Glycine SDS Sample Buffer and Novex Tris-Glycine SDS Running Buffer. To separate native proteins use Novex Tris-Glycine Native Sample Buffer and Novex Tris-Glycine Native Running Buffer.

See all available buffers and reagents available for SDS-PAGE

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialRunning Buffer
Recommended StorageIncludes Novex™ Tris-Glycine SDS Running Buffer (10X, 5 L)
Concentration10X
For Use With (Equipment)Novex Tris-Glycine Gels
Physical FormLiquid
Product LineNovex
Quantity5 L
Unit SizeEach

Frequently asked questions (FAQs)

Where do I find buffer recipes for your precast protein gels?

The formulations of buffers for our precast protein gels can be found at this link: https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-electrophoresis-buffers-reagents.html

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

How should I store Novex Tris-Glycine SDS Running Buffer (10X)?

We recommend storing Novex Tris-Glycine SDS Running Buffer (10X) at 4 to 25 degrees C.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.