MagicMark™ XP Western Protein Standard
Invitrogen™

MagicMark™ XP Western Protein Standard

MagicMark XP Western Protein Standard consists of nine recombinant proteins (20–220 kDa), each of which contains an IgG binding site.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LC5602250 μL
LC5602X44 x 250 μL
LC560350 μL
Catalog number LC5602
Price (KRW)
496,000
Online offer
Ends: 30-Sep-2026
566,000
Save 70,000 (12%)
Each
Quantity:
250 μL
Request bulk or custom format
Price (KRW)
496,000
Online offer
Ends: 30-Sep-2026
566,000
Save 70,000 (12%)
Each
MagicMark XP Western Protein Standard consists of nine recombinant proteins (20–220 kDa), each of which contains an IgG binding site. The IgG binding site binds the primary or secondary antibody used for detection of the target protein, allowing direct visualization of the standard on the western blot. The MagicMark XP Standard is compatible with most western kits and substrates (chemiluminescent, chromogenic, and fluorescent). The protein standard is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Compare and view all other protein standards and ladders ›

Applications
• Western blotting: detection of the nine unstained bands via the detection method used for the target protein. Compatible with chemiluminescent substrates and fluorescent secondary antibodies (not recommended for antibodies labeled with fluors in the 500–550 nm channel).
• Protein size estimation: protein sizing on SDS-polyacrylamide gels and on western blot using western blot detection method
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodUser defined detection system
Gel CompatibilityBolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Molecular Weight (g/mol)220, 120, 100, 80, 60, 50, 40, 30, 20 kDa
Quantity250 μL
Ready to LoadYes
Shipping ConditionDry Ice
Number of Markers9
Product LineMagicMark
Product TypeProtein Ladder
Size Range20 to 220 kDa
Stain TypeUnstained
System TypeWestern Blotting, SDS-PAGE
Unit SizeEach
Contents & Storage
250 μL of ready-to-use standard mixture is provided in a plastic vial. Loading buffer consists of 125 mM Tris-HCl (pH 6.8), 10 mM DTT, 17.4% Glycerol, 3% SDS, and 0.025% Bromophenol Blue.

Store at -20°C.

Frequently asked questions (FAQs)

What can I use as a positive control or marker on my western blots?

The Positope control protein (Cat. No. R90050) is designed to be used as a positive control in western blotting for a variety of antibodies. The control protein is a 53 kDa highly purified recombinant protein that contains seven epitope tags including the Xpress, c-myc, V5, His(C-term), HisG(N-term), Thioredoxin, and GFP.

The MagicMark XP Western Protein Standard (Cat. No. LC5602) consists of peptides that contain an IgG binding domain. These peptides can be detected with your regular antibodies and provide a way for you to get good molecular weight estimations without having to rely on transferred pre-stained standards.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What is the composition of the MagicMark XP Western Protein Standard?

MagicMark XP standards consist of a mixture of eight E. coli-expressed recombinant proteins ranging in size from 20 to 120 kDa. Each protein contains an IgG binding sequence from Protein G. This IgG binding site binds the primary or secondary antibody used to detect your target protein, allowing you to detect the MagicMark standard directly.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I used your MagicMark XP Western Protein Standard and obtained a very weak or almost no signal after the western detection. Can you please help me troubleshoot?

Here are some suggestions:

- Verify that the detection reagents are working well. Optimize the antibody concentration to obtain best results.
- Make sure that the amount of standard loaded on the gel is correct.
- Optimize the transfer conditions (current, voltage, transfer time).
- Enzyme-conjugated primary antibodies may not bind efficiently with the proteins in the MagicMark XP Western Protein Standard. We recommend using unconjugated primary antibody, followed by the addition of enzyme-conjugated secondary antibody.
Note: The Anti-myc-AP/HRP and Anti-V5-AP/HRP antibodies do not bind to MagicMark XP proteins.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I used one of your protein standards for a western transfer and noticed that some of the lower-molecular weight protein bands passed through the membrane. How can I resolve this issue?

- Decrease voltage, current or length of transfer time
- Make sure that the methanol concentration in the transfer buffer is proper; use a methanol concentration of 10-20% methanol removes the SDS from SDS-protein complexes and improves the binding of protein to the membrane.
- Make sure that the SDS concentration (if added) in the transfer buffer is proper, don't use more than 0.02-0.04% SDS. Using too much SDS can prevent binding of proteins to the membrane.
- Check the pore size of the membrane and the size of the target protein. Proteins smaller than 10 kDa will easily pass through a 0.45 µm pore size membrane. If proteins smaller than 10 kDa are of interest, it would be better to use a 0.2 µm pore size membrane.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I used one of your protein standards for a western transfer and noticed that some of the higher-molecular weight bands transferred very poorly to the membrane. Can you offer some tips?

- Increase voltage, current or length of time for transfer
- SDS in the gel and in the SDS-protein complexes promotes elution of the protein from the gels but inhibits binding of the protein to membranes. This inhibition is higher for nitrocellulose than for PVDF. For proteins that are difficult to elute from the gel such as large molecular weight proteins, a small amount of SDS may be added to the transfer buffer to improve transfer. We recommend pre-equilibrating the gel in 2X Transfer buffer (without methanol) containing 0.02-0.04% SDS for 10 minutes before assembling the sandwich and then transferring using 1X transfer buffer containing 10% methanol and 0.01%SDS.
- Methanol removes the SDS from SDS-protein complexes and improves the binding of protein to the membrane, but has some negative effects on the gel itself, leading to a decrease in transfer efficiency. It may cause a reduction in pore size, precipitation of some proteins, and some basic proteins to become positively charged or neutral. Make sure that the methanol concentration in the transfer buffer is not more than 10-20% and that high-quality, analytical grade methanol is used.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.