The Mark12 Unstained Standard is suitable for NuPAGE, Tris-Glycine and Tricine gels. Here are the molecular weights and origins of the proteins in the Mark12 Unstained Standard:
- Myosin: 200 kDa; Rabbit muscle
- b-galactosidase: 116.3kDa; E. coli
- Phosphorylase B: 97.4 kDa; Rabbit muscle
- Bovine serum albumin: 66.3 kDa; Bovine serum
- Glutamic dehydrogenase: 55.4 kDa; Bovine liver
- Lactate dehydrogenase: 36.5 kDa; Porcine
- Carbonic anhydrase: 31.0 kDa; Bovine
- Trypsin inhibitor: 21.5 kDa; Soybean
- Lysozyme: 14.4 kDa; Egg white
- Aprotinin: 6.0 kDa; Bovine lung
- Insulin B chain: 3.5* kDa; Bovine pancreas
- Insulin A chain: 2.5*kDa; Bovine pancreas
Note: The 3.5 kDa and 2.5 kDa bands are visible only on NuPAGE Bis-Tris gels run using 1X MES SDS Running buffer.
Find additional tips, troubleshooting help, and resources within our
Protein Assays and Analysis Support Center.