Here are possible causes and solutions:
- Improper labeling. Make sure that the labeling protocol is correctly followed to obtain the best results. Make sure you have added the Lumio Green Detection Reagent to the samples prior to electrophoresis. Limit exposure of the Lumio Gel Sample Buffer (4X) to air. Always return the Lumio Green Reagent and Lumio Enhancer to ?20 degrees C immediately after use to preserve the activity of buffers.
- Low protein load or low expression level. Check total protein loaded on the gel by staining the gel with a total protein stain as described in the manual. Load at least 1 pmole of the Lumio fusion protein. Make sure the Lumio tag is in frame and the protein is expressed properly. A positive control is supplied with the Lumio vectors to verify the expression protocol.
- The gel is exposed to UV light for a long time. The fluorescent dye of the Lumio Green Reagent is sensitive to photobleaching, so avoid exposing the gel to UV light for a long time.
- The gel is not visualized immediately or imaged properly. Be sure to visualize the gel after removing the gel from the cassette and view the gel immediately after electrophoresis. Use a UV transilluminator or a laser-based scanner using appropriate filters as described in the manual.
Tip: If you have run BenchMark Fluorescent Protein Standard on the same gel and can view the standard bands on the gel, then you are imaging the gel properly.
Find additional tips, troubleshooting help, and resources within our
Protein Assays and Analysis Support Center.