Orion Lab Star PH111 Bench pH Meters
Orion Lab Star PH111 Bench pH Meters
Orion Lab Star PH111 Bench pH Meters
Orion Lab Star PH111 Bench pH Meters
Orion Lab Star PH111 Bench pH Meters
Thermo Scientific™

Orion Lab Star PH111 Bench pH Meters

Orion Lab Star PH111 benchtop pH meters measure pH or ORP with mV and temperature, feature a large backlit display for easy viewability, and offer small footprint to save bench space. The Orion Lab Star pH meter is ideal for simplified, budget-friendly lab testing and data recording.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipo
LSTAR1116Kit biológico de medición de pH/mV
LSTAR1110Medidor de pH/mV con soporte
LSTAR1115Kit estándar de medición de pH/mV
LSTAR1117Medidor de pH/mV bajo principal Kit biológico
LSTAR1118Kit de muestras difíciles de medición de pH/mV
Número de catálogo LSTAR1116
Precio (MXN)
-
Tipo:
Kit biológico de medición de pH/mV
Discover the award-winning Orion Lab Star PH111 pH/mV bench meter. Measure pH or ORP simultaneously with mV and temperature. Featuring a large, backlit display for easy viewability and small footprint to save bench space, this pH meter is ideal for simplified, budget-friendly lab testing. Use the onscreen stabilizing and ready indicators to help identify when pH meter and ORP meter readings are complete. View active pH, ORP and temperature calibration logs with date and time stamp, record key measurements using the 500-point data log with date and time stamp, and output all logs to a printer or computer.

Orion Lab Star PH111 pH Meter Features

  • Budget-friendly pH meter for simplified pH and ORP testing
  • Display shows pH, mV and temperature simultaneously for accurate data recording
  • Measure -2.000 to 18.000 pH, -2000.0 to +2000.0 mV and -5.0 to 105.0 °C or 23.0 to 221.0 °F to ensure comprehensive measurement capabilities
  • Perform one to five point pH calibrations, one point ORP calibrations and one point temperature adjustment calibrations for proficient SOP compliance
  • Large 5 inch backlit display makes viewing and recording measurements easy
  • Small meter footprint saves valuable bench space in your lab
  • Onscreen stabilizing and ready indicators help identify when readings are complete
  • pH electrode condition icon helps ensure reliable electrode performance
  • Date and time data stamped data and calibration logs support GLP compliance
  • View active pH, ORP and temperature calibration logs with date and time stamp to ensure accurate, reproducible measurements
  • Use 500-point data log with date and time stamp to record key measurements
  • Output data and calibration logs to a printer or computer for long-term data storage

Meter and Meter Kits

  • LSTAR1110 is the pH meter and stand option, which allows you to purchase your pH electrode separately
  • LSTAR1115 is the standard pH meter kit option, for common pH samples such as drinking water, tap water, buffers (non-TRIS) and general purpose student use
  • LSTAR1116 is the easy-to-clean bio pH meter kit option, for pH samples containing TRIS buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; included refillable pH electrode is easy-to-clean and restore
  • LSTAR1117 is the low-maintenance bio pH meter kit option, for pH samples containing TRIS buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; included gel-filled pH electrode is low-maintenance and easy to operate
  • LSTAR1118 is the difficult sample pH meter kit option, for pH samples that are thick, viscous or semi-solid including foods, beverages, wastewaters, emulsions, soils, slurries, suspended solids or sludges
Especificaciones
Adaptador de CAFuente de alimentación universal de 100-240 V 50-60 Hz con conexión de medidor de giro y bloqueo y enchufes de pared intercambiables para tomacorrientes de EE. UU./Japón, Europa, Reino Unido/Singapur, Australia/Nueva Zelanda y China
Exactitud (temperatura)±0,3 °C/±0,5 °F
Exactitud (mV)±0,2 o ±0,05 %, el que sea mayor
Exactitud (pH)±0,005, ± ± 1 LSD
Salida de alarmaAlarma de recordatorio de calibración
Puntos de calibración (pH)1 a 5 puntos
Certificaciones/conformidadCE, TUV 3-1, FCC Clase A
Detalles de gestión de datosTransferencia de todos los puntos de datos o de puntos de datos individuales a la impresora u ordenador
DescripciónKit biológico de medidor de sobremesa de pH/mV Orion Lab Star PH111 fácil de limpiar
Dimensiones (Largo x Ancho x Alto)198,3 x 155,2 x 61,7 mm
Para utilizar con (aplicación)Opción de kit de medidor de pH biológico fácil de limpiar, para muestras de pH que contienen tampones TRIS, proteínas, sulfuros, haluros u otros componentes que pueden interferir con el cloruro de plata; el electrodo de pH recargable incluido es fácil de limpiar y restaurar
Clasificación IPIP54
IncluyeElectrodo de pH de cuerpo epoxi recargable de doble unión GD9156BNWP, sonda de temperatura ATC de acero inoxidable 927007MD, 4/7/10 tampón de pH 916099, kit de solución de almacenamiento y limpieza de 60 ml, medidor con portaelectrodos, cable para PC, adaptador de corriente universal
EntradasBNC para pH o ORP, MiniDIN de 8 pines para ATC
MemoriaNo volátil
SalidasOrdenador o impresora (STARA-106)
Intervalo (ORP)±2000,0 mV con desviación de ±250 mV
Resolución (temperatura)0,1 °C (0,1 °F)
Resolución (mV)0,1 mV
Calibración de la sonda de temperaturaCalibración de desviación de la temperatura de 1 punto
Intervalo de temperatura (imperial)23,0° hasta 221,0°F
Selección de temperaturaManual o automático con sonda ATC de temperatura
TipoKit biológico de medición de pH/mV
GarantíaGarantía del medidor de 3 años
Peso (imperial)1,54 lb
Peso (métrico)700 gramos
Backlight OptionSí
CalibraciónpH, ORP y temperatura
Calibration Points1 (ORP)
Puntos de datos500 con sello de fecha y hora
Tipo de pantallaLCD, 12,7 cm (5 pulg) retroiluminada
Tipo de función de registroAutomático con modos de lectura automática y temporizada, manual con modo de lectura continuo
Measurement ModespH con mV y temperatura o ORP con mV y temperatura, ORP with mV and Temperature
Intervalo (mV)±2000,0
Intervalo (pH)Desde -2,000 hasta +18,000
Resolución (pH)0,001, 0,01, 0,1
Intervalo de temperatura (métrico)De -5,0 °C a +105,0 °C
Unit SizeEach

Preguntas frecuentes

How long is the warranty period for Orion pH meters?

At Thermo Fisher Scientific, we stand behind the quality and reliability of our instruments. That's why we offer a minimum two-year (2) warranty on all Orion benchtop and portable pH meters. Obtain peace of mind knowing that your investment is protected. Should any issues arise within the warranty period, our team of experts is committed to providing you with prompt and efficient service. Be sure to check instrument specifications to confirm the warranty coverage period.

How do I perform a calibration using Orion pH meters?

Calibration is an important step to obtaining accurate results on your Orion benchtop or portable pH meter. Before beginning the calibration process, ensure that you are utilizing freshly poured, non-expired calibration buffers to help ensure accurate results. Follow the steps below to calibrate your pH meter.

• Prepare the calibration solutions: Obtain at least two pH reference solutions with known pH values. Ideally, use one solution close to the expected pH of your sample, and another solution at a different pH value. Ensure that the solutions are fresh and have not expired.
• Clean the electrode: Rinse the electrode with distilled or deionized water to remove any debris or contaminants. If necessary, gently wipe the electrode with a soft cloth or tissue.
• Turn on the pH meter: Power on the pH meter and allow it to warm up, if needed.
• Immerse the electrode: Immerse the electrode into a beaker containing the first calibration solution. Ensure that the electrode is fully submerged and that it is not touching the sides or bottom of the beaker.
• Allow stabilization: Allow the pH reading to stabilize. This may take a few moments, depending on the specific meter and solution being used. The reading should become steady and remain constant.
• Adjust the pH value: Using the calibration controls on the pH meter, adjust the reading to match the known pH value of the calibration solution. Reference the user manual for specific guidance on adjusting the pH.
• Rinse the electrode: After calibration, rinse the electrode with distilled or deionized water to remove any residual calibration solution.
• Repeat calibration: Repeat the calibration steps with the second calibration solution, following the same process as before.
• Verify accuracy: After calibration, it is recommended to check the pH meter's accuracy by measuring a pH solution of known value. This can help ensure the reliability of your measurements.

To learn more about pH meter calibration, download our technical note: Understanding Calibration of Laboratory pH Systems. See a live calibration by watching our video: Understanding Calibration of Laboratory pH Systems.

How do I perform an analysis using Orion pH meters?

Before beginning an analysis on your Orion benchtop or portable pH meter, assemble your materials. You will need a pH electrode suitable for your application, calibration buffer solutions, and a test sample. Follow the steps below to obtain your pH analysis.

• Calibrate the pH meter: Start by calibrating your pH meter using the calibration buffer solutions. Typically, you will need pH 4, pH 7, and pH 10. Be sure to reference the user manual for instructions on how to properly calibrate your pH meter.
• Prepare your sample: Ensure that your sample is at the desired temperature (most frequently room temperature). If necessary, filter the sample to remove any particles or impurities that could impact the pH reading.
• Immerse the electrode: Dip the pH electrode into the sample. Avoid touching the sides or bottom of the container to prevent inaccurate readings.
• Allow for stabilization: Give the pH meter some time to stabilize and reach a steady reading. This may take a few seconds to a minute depending on your meter and the characteristics of the sample.
• Record the pH value: Once the pH meter stabilizes, record the pH value displayed on the meter. If applicable, also record the temperature value.
• Rinse the electrode: After each measurement, rinse the electrode with distilled or deionized water to remove any residue or contaminants. This helps ensure accurate subsequent readings.
• Repeat (if necessary): If you need to analyze multiple samples, repeat the process with each sample, recalibrating the pH meter if required.

To learn more about how to perform a pH analysis, watch our video:Measuring Drinking Water Using the Orion Versa Star Pro Bench Meter

How do I choose an Orion pH electrode for my application?

When choosing an Orion pH electrode for your application there are many factors to consider. Sample type, container dimensions, accuracy/ precision requirements, junction design, durability, maintenance, and meter compatibility are all key features to evaluate.
To learn more about how to choose the right electrode for your application, watch our videos: How to Choose the Right pH Electrode and Understanding Your Sample to Select the Right pH Electrode

What supplies do I need to perform an analysis with Orion pH meters?

To perform an analysis using Orion benchtop and portable pH meters, you will need the following supplies: pH electrode (or micro electrode for small sample volumes/ containers), calibration buffer solutions, distilled or deionized water, sample containers, stirrer probe (optional), and electrode cleaning solution(s) (s). For your convenience, our all-in-one pH buffer kits can be purchased, which include pH buffers (4.01 pH, 7.00 pH, and 10.01 pH), electrode cleaning solution, and electrode and storage solution.