Lykos PAT Software – Pro
Thermo Scientific™
Lykos PAT Software – Pro
Lykos PAT Software Pro supports users of the MarqMetrix All-In-One Process Raman Analyzer by connecting instruments across workflows while sharing key data, including chemometric predictions.
Have Questions?
카탈로그 번호 LYKOSPRO
제품 가격(KRW)
-
Enable Open Platform Communication Unified Architecture (OPC™ UA) integration of the Thermo Scientific™ MarqMetrix™ All-In-One Process Raman Analyzer chemometric results with Lykos PAT Software – Pro. Lykos PAT Software – Pro improves processes with an intuitive system designed to help manage and integrate instruments across workflows, streamline data management through digital connectivity, and more.
Key Features:
Lykos PAT Software – Pro OPC–UA allows Lykos users to:
  • Connect multiple instruments within the workflow (machine-to-machine communication)
  • Pass chemometric predictions, in real-time, to an OPC client
  • Streamline results into another software system
  • Use results in an automated feedback loop
Compliance Add-On:
Lykos also supports cGMP & 21 CFR Compliance. Visit the Lykos PAT Software – Compliance to learn more.
Qualification Services:
Our service and support team offers IQ/OQ qualification services for your instrument and software.
사양
설명Lykos PAT Software - Pro
용도(장비)MarqMetrix All-In-One Process Raman Analyzer
제품 유형Software License
Unit SizeEach