In the MitoProbe TMRM Assay Kit for Flow Cytometry, should CCCP be present when adding the TMRM?
Yes. The CCCP should be first added to the cell sample, incubated for 5 mins, and then the TMRM should be added and incubated for another 30 mins.