MitoProbe™ Transition Pore Assay Kit, for flow cytometry
Invitrogen™

MitoProbe™ Transition Pore Assay Kit, for flow cytometry

The MitoProbe™ Transition Pore Assay Kit provides a proven method (Petronilli V. et al. 1989, 1999) of measuring mitochondrial permeabilityRead more
Have Questions?
Catalog NumberQuantity
M341531 kit
Catalog number M34153
Price (USD)
527.65
Online exclusive
704.00
Save 176.35
Each
Quantity:
1 kit
Price (USD)
527.65
Online exclusive
704.00
Save 176.35
Each
The MitoProbe™ Transition Pore Assay Kit provides a proven method (Petronilli V. et al. 1989, 1999) of measuring mitochondrial permeability transition pore opening. The kit includes a dye and a quencher that are easily loaded into cells. In healthy cells, the dye, but not the quencher, enters mitochondria. The mitochondria remain brightly fluorescent until mitochondrial pore activation permits quenching of the fluorescence.

View a selection guide for all apoptosis assays for flow cytometry.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Excitation/Emission488
No. of Reactions100 Reactions
Product LineMitoProbe
Product TypeTransition Pore Assay Kit
Quantity1 kit
Shipping ConditionRoom Temperature
ConjugateCalcein AM
FormatTube
Unit SizeEach
Contents & Storage
Contains 5 vials of calcein, AM (50 μg of dry powder/vial), 1 vial of cobalt (II) Chloride hexahydrate (1.2 mL, 80 mM in saline), 1 vial of ionomycin (55 μg), calcium salt, and 1 vial of DMSO (1.5 mL).

Store in freezer (-5 to -30°C).

Frequently asked questions (FAQs)

What are the advantages of flow cytometry?

-Measures data from single cells.
-Data are obtained for a large number of cells, generating a rich statistical analysis of cell populations.
-Because single cells are measured, it will reveal heterogeneity within a population.
-With the ability to multiplex, small sub-populations can be identified.
-Thousands of cells can be analyzed rapidly.
-It is ideally suited for blood samples and other cells in suspension.
-Data can be re-analyzed multiple times after acquisition.
-Flow cytometry files (FCS) can be archived.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What kinds of applications can I run on a flow cytometer?

There are several applications, some of which include immunophenotyping, cell cycle analysis, apoptosis assays such as annexin V staining, CellEvent Caspase-3/7 assay, and TUNEL assay, cell viability, proliferation assays such as CellTrace assay and Click-iT EdU assay, measurements of mitochondrial potential with MitoProbe assays, and cell counting using counting beads.

Find additional tips, troubleshooting help, and resources within our Flow Cytometry Support Center.

Citations & References (16)

Citations & References
Abstract
Cysteine 203 of cyclophilin D is critical for cyclophilin D activation of the mitochondrial permeability transition pore.
Authors:Nguyen TT, Stevens MV, Kohr M, Steenbergen C, Sack MN, Murphy E,
Journal:J Biol Chem
PubMed ID:21930693
The mitochondrial permeability transition pore (mPTP) opening plays a critical role in mediating cell death during ischemia/reperfusion (I/R) injury. Our previous studies have shown that cysteine 203 of cyclophilin D (CypD), a critical mPTP mediator, undergoes protein S-nitrosylation (SNO). To investigate the role of cysteine 203 in mPTP activation, we ... More
Transient and long-lasting openings of the mitochondrial permeability transition pore can be monitored directly in intact cells by changes in mitochondrial calcein fluorescence.
Authors:Petronilli V, Miotto G, Canton M, Brini M, Colonna R, Bernardi P, Di Lisa F
Journal:Biophys J
PubMed ID:9929477
'The occurrence and the mode of opening of the mitochondrial permeability transition pore (MTP) were investigated directly in intact cells by monitoring the fluorescence of mitochondrial entrapped calcein. When MH1C1 cells and hepatocytes were loaded with calcein AM, calcein was also present within mitochondria, because (i) its mitochondrial signal was ... More
Calpain and caspase orchestrated death signal to accomplish apoptosis induced by resveratrol and its novel analog hydroxystilbene-1 [correction of hydroxstilbene-1] in cancer cells.
Authors:Guha P, Dey A, Dhyani MV, Sen R, Chatterjee M, Chattopadhyay S, Bandyopadhyay SK,
Journal:J Pharmacol Exp Ther
PubMed ID:20484155
'Stomach ulceration is a major side effect of most chemopreventive drugs. We have established that although resveratrol is a promising chemopreventive compound, it delays the ulcer healing process. However, its analog hydroxystilbene-1 (HST-1) was devoid of such an ulcerogenic side effect. Consequently, here we tried to explore the chemopreventive efficacy ... More
Secondary coenzyme Q10 deficiency triggers mitochondria degradation by mitophagy in MELAS fibroblasts.
Authors:Cotán D, Cordero MD, Garrido-Maraver J, Oropesa-Ávila M, Rodríguez-Hernández A, Gómez Izquierdo L, De la Mata M, De Miguel M, Lorite JB, Infante ER, Jackson S, Navas P, Sánchez-Alcázar JA,
Journal:FASEB J
PubMed ID:21551238
Mitochondrial encephalomyopathy, lactic acidosis, and stroke-like episodes (MELAS) is a mitochondrial disease most usually caused by point mutations in tRNA genes encoded by mtDNA. Here, we report on how this mutation affects mitochondrial function in primary fibroblast cultures established from 2 patients with MELAS who harbored the A3243G mutation. Both ... More
Imaging the mitochondrial permeability transition pore in intact cells.
Authors:Petronilli V, Miotto G, Canton M, Colonna R, Bernardi P, Di Lisa F
Journal:Biofactors
PubMed ID:9914828
The involvement of mitochondrial permeability transition pore (MTP) in cellular processes is generally investigated by indirect means, such as changes in mitochondrial membrane potential or pharmacological inhibition. However, such effects could not be related univocally to MTP. In addition, source of errors could be represented by the increased retention of ... More