Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 5
This antibody reacts with human platelets.
CD41 is a transmembrane glycoprotein (integrin family) composed of two chains GPIIb alpha (heavy chain; 120 kDa) and GPIIb beta (light chain; 23 kDa). CD61 non-covalently associates with CD41.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alphaIIb protein; antigen CD61; CD41; CD61; glycoprotein IIIa precursor; GPalpha IIb; GPIIb; GPIIIa; integrin alpha 2b; Integrin alpha-2b; Integrin alpha-IIb; integrin beta 3; integrin beta 3 subunit; integrin beta chain, beta 3; Integrin beta-3; integrin beta-3 subunit; integrin homolog; integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41); integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); ITGA2B-like protein; plate glycoprotein IIIa (GPIIIa); platelet fibrinogen receptor, alpha subunit; platelet glycoprotein IIb (GPIIb); platelet glycoprotein IIb of IIb/IIIa complex; Platelet membrane glycoprotein IIb; Platelet membrane glycoprotein IIIa; platelet-specific antigen BAK; protein phosphatase 1, regulatory subunit 93; unnamed protein product
Gene Aliases: BDPLT16; BDPLT2; BDPLT24; CD41; CD41B; CD61; GP2B; GP3A; GPIIb; GPIIIa; GT; GT1; GT2; GTA; HPA3; INTB3; ITGA2B; ITGAB; ITGB3; PPP1R93
UniProt ID: (Human) P08514, (Human) P05106
Entrez Gene ID: (Bovine) 515011, (Human) 3674, (Human) 3690, (Bovine) 282642
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support