Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • PSD-95 Antibodies

          Invitrogen

          PSD-95 Monoclonal Antibody (7E3-1B8)

          Advanced Verification
          75 Published Figures
          329 References
          View all (51) PSD-95 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PSD-95 Monoclonal Antibody (7E3-1B8)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (10)
          • Published Figures (75)
          • Advanced Verification (2)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 87

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PSD-95 Antibody (MA1-046) in ICC/IF

          Immunofluorescent analysis of PSD95 using Anti-PSD95 Monoclonal Antibody (7E3-1B8) (Product # MA1-046) shows staining in Hela Cells. PSD95 staining (green), F-Actin staining with Phalloidin (red) and nuclei with DAPI (blue) is shown. Cells were grown on chamber slides and fixed with formaldehyde prior to staining. Cells were probed without (control) or with or an antibody recognizing PSD95 (Product # MA1-046) at a dilution of 1:20 over night at 4°C, washed with PBS and incubated with a DyLight-488 conjugated secondary antibody (Product # 3... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Flow Cytometry (Flow)
          PSD-95 Antibody in Flow Cytometry (Flow)
          PSD-95 Antibody in Flow Cytometry (Flow)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot, Immunomicroscopy (WB, IM)
          PSD-95 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Immunomicroscopy (IM)
          PSD-95 Antibody in Immunomicroscopy (IM)
          PSD-95 Antibody in Immunomicroscopy (IM)
          PSD-95 Antibody in Immunomicroscopy (IM)
          PSD-95 Antibody
          PSD-95 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA1-046

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000
          View 103 publications 103 publications

          Immunohistochemistry (IHC)

          -
          View 83 publications 83 publications

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:500
          View 118 publications 118 publications

          Flow Cytometry (Flow)

          2 µg/test
          -

          Immunoprecipitation (IP)

          Assay-dependent
          View 12 publications 12 publications

          Neutralization (Neu)

          -
          View 1 publication 1 publication

          Functional Assay (Functional)

          -
          View 1 publication 1 publication

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          Immunomicroscopy (IM)

          -
          View 4 publications 4 publications

          Miscellaneous PubMed (Misc)

          -
          View 5 publications 5 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat, Xenopus

          Published species

          Avian, Bovine, Chicken, Human, Invertebrate, Marsupial, Mouse, Non-human primate, Rabbit, Rat, Zebrafish

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          7E3-1B8

          Immunogen

          Purified recombinant rat PSD-95.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2092361

          Product Specific Information

          MA1-046 detects post synaptic density 95 (PSD-95) from human, rat, xenopus laevis and mouse tissues.

          MA1-046 has been successfully used in Western blot, immunofluorescence, immunocytochemistry, immunohistochemistry, flow cytometry and immunoprecipitation procedures. By Western blot, this antibody detects an ~95 kDa protein representing PSD-95 from rat brain extracts. Immunofluorescence staining of PSD-95 in rat hippocampal cells with MA1-046 yields a staining pattern coincident with NMDA receptor.

          In immunofluorescence staining, fixation with cold methanol is recommended.

          The MA1-046 immunogen is purified recombinant rat PSD-95.

          This antibody was orginally validated as part of a Thermo Scientific Cellomics High Content Screening Kit. The antibody sold separately may have slightly different performance and may need to be further optimized for the best results.

          Target Information

          Post Synaptic Density 95 kDa (PSD-95), also known as synapse associated protein 90 kDa (SAP90), is one of a family of membrane-associated proteins found in the postsynaptic density in forebrain neurons and certain presynaptic structures in the cerebellum. Like other members of the family, PSD-95 has three 90 amino acid repeats called PDZ domains followed by an SH3 domain and a yeast guanylate kinase homology (GuK) domain. PSD-95 is believed to participate in the clustering of certain proteins, including NMDA receptors, Shaker-type potassium channels at the synaptic membrane in central nervous system (CNS) neurons. There are two principal modes of interaction between PSD-95 and other proteins. NMDA receptors and shaker-type potassium channels both share C-terminal sequence homology consisting of a threonine/serine-X-valine-COOH (T/SXV) motif. Other neuronal proteins that share this motif (beta 1 adrenergic receptor, some serotonin receptors, some sodium channel subunits, and additional potassium channel subunits), and some of these proteins may interact with PSD-95 by binding to its PDZ domains. Neuronal nitric oxide synthase (nNOS), which lacks the T/SXV motif but which has its own PDZ domain, has been shown to associate with PSD-95 in vitro through a pseudo-homotypic PDZ-PDZ interaction.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: discs large homolog 4; discs, large homolog 4; FLJ97752; FLJ98574; post synaptic density; post-synaptic density protein 95; postsynaptic density protein 95; PSD-95; PSD-95 alpha 2b; PSD-95 beta; SAP-90; synapse-associated protein 90; synapse-associated protein SAP90; Tax interaction protein 15; unnamed protein product

          View more View less

          Gene Aliases: Dlgh4; MRD62; PSD-95; PSD95; SAP-90; SAP90; SAP90A

          View more View less

          UniProt ID: (Human) P78352, (Rat) P31016, (Mouse) Q62108

          View more View less

          Entrez Gene ID: (Human) 1742, (Rat) 29495, (Mouse) 13385, (Western clawed frog) 779559

          View more View less

          Function(s)
          protein binding kinase binding protein kinase binding protein phosphatase binding GKAP/Homer scaffold activity PDZ domain binding protein binding, bridging beta-1 adrenergic receptor binding D1 dopamine receptor binding P2Y1 nucleotide receptor binding acetylcholine receptor binding ionotropic glutamate receptor binding macromolecular complex binding neuroligin family protein binding scaffold protein binding receptor binding frizzled binding kinesin binding beta-2 adrenergic receptor binding glutamate receptor binding structural constituent of postsynaptic density structural molecule activity neurexin family protein binding scaffold/adaptor protein
          Process(es)
          negative regulation of receptor internalization signal transduction positive regulation of cytosolic calcium ion concentration chemical synaptic transmission nervous system development learning synaptic vesicle maturation social behavior protein localization to synapse locomotory exploration behavior cellular response to potassium ion establishment of protein localization establishment or maintenance of epithelial cell apical/basal polarity regulation of long-term neuronal synaptic plasticity positive regulation of synaptic transmission neuromuscular process controlling balance long term synaptic depression dendritic spine morphogenesis protein-containing complex assembly vocalization behavior AMPA glutamate receptor clustering receptor localization to synapse cell-cell adhesion NMDA selective glutamate receptor signaling pathway regulation of postsynaptic membrane neurotransmitter receptor levels neurotransmitter receptor localization to postsynaptic specialization membrane positive regulation of neuron projection arborization positive regulation of excitatory postsynaptic potential regulation of grooming behavior developmental maturation cerebral cortex development neuronal ion channel clustering regulation of neuronal synaptic plasticity positive regulation of dendrite morphogenesis positive regulation of synapse assembly protein localization to membrane dendritic spine organization maintenance of postsynaptic density structure positive regulation of AMPA glutamate receptor clustering regulation of N-methyl-D-aspartate selective glutamate receptor activity locomotory behavior response to cocaine
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-l8rg4:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0