Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
MA1-10041 recognizes phosphorylated NF-H KSP (lysine-serine-proline) type sequences.
Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-H has an important role in mature axons and is involved in amyotrophic lateral sclerosis (ALS).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 200 kDa neurofilament protein; heavy neurofilament polypeptide; heavy neurofilament subunit; high molecular weight neurofilament; hSTEp; NEF3; neurofilament; neurofilament 200kDa; neurofilament heavy polypeptide; Neurofilament triplet H protein; neurofilament, heavy polypeptide; neurofilament, heavy polypeptide 200kDa; NF 200 kD; NF-H; Nfh; NF200; NHC; unnamed protein product
Gene Aliases: CMT2CC; mKIAA0845; NF-H; NF200; NFH
UniProt ID: (Human) P12036, (Mouse) P19246
Entrez Gene ID: (Horse) 100064328, (Human) 4744, (Pig) 100156492, (Chicken) 417020, (Mouse) 380684, (Rat) 24587
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support