Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
MA1-10624 detects Chymotrypsin from human samples.
MA1-10624 has been successfully used in ELISA and Western blot applications.
The MA1-10624 immunogen is purified human pancreatic Chymotrypsin.
This gene encodes a member of the peptidase S1 family. The encoded protein is a serum calcium-decreasing factor that has chymotrypsin-like protease activity. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: caldecrin; chymotrypsinogen C; elastase 4; elastase IV; serum calcium decreasing factor; serum calcium-decreasing factor; unnamed protein product
Gene Aliases: CLCR; ELA4
UniProt ID: (Human) Q99895
Entrez Gene ID: (Human) 11330
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support