Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody is a monoclonal mixture of FITC-conjugated CD10 (clone RXB.14, isotype IgG1) and PE-conjugated CD19 (clone 1G9, isotype IgG1).
Anti-hCD10 recognizes a human common acute lymphoblastic antigen (CALLA), MW 100 kDa. The CD10 antigen is identical to human membrane-associated neutral endopeptidase (NEP;EC 3.3.24.11), also known as enkephalinase. The CD10 antigen is found on lymphocytes from patients with acute B-lymphoid leukemia. The antigen is also present on a wide variety of normal and neoplastic cell types including renal epithellum, fibroblasts, granulocytes and some lymphoma, melanoma and glioma cell lines. Identification of CD19 PE human B cells associated approximately 10% of peripheral blood lymphocytes expressing the 95,000 M.W. surface antigen.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: atriopeptidase; AW495831; B-lymphocyte surface antigen B4; CD19 differentiation antigen; common acute lymphoblastic leukemia antigen precursor; common acute lymphocytic leukemia antigen; differentiation antigen CD19; membrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, CD10); NEP-390; NEP-411; neprilysin-390; neprilysin-411; neutral endopeptidase 24.11; skin fibroblast elastase; T-cell surface antigen Leu-12; unnamed protein product
Gene Aliases: B4; CALLA; CD10; CMT2T; CVID3; NEP; SCA43; SFE
UniProt ID: (Human) P15391, (Human) P08473
Entrez Gene ID: (Human) 930, (Human) 4311
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support