Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Active/Pro-Caspase 3 Antibodies

          Invitrogen

          Active/Pro-Caspase 3 Monoclonal Antibody (CPP32 4-1-18)

          Advanced Verification
          8 Published Figures
          19 References
          View all (5) Active/Pro-Caspase 3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Active/Pro-Caspase 3 Monoclonal Antibody (CPP32 4-1-18)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (7)
          • Published Figures (8)
          • Advanced Verification (3)
          Active/Pro-Caspase 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Active/Pro-Caspase 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 18

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Active/Pro-Caspase 3 Antibody (MA1-16843) in IHC (P)

          Immunohistochemistry analysis of Active/Pro-Caspase 3 in immersion fixed paraffin-embedded sections of human bladder. Samples were incubated with Active/Pro-Caspase 3 monoclonal antibody (Product # MA1-16843) using a dilution of 1:300 for 1 hour at room temperature followed by anti-mouse IgG VisUCyte HRP polymer. Tissue was stained using DAB (brown) and counterstained with hematoxylin (blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Active/Pro-Caspase 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Active/Pro-Caspase 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Active/Pro-Caspase 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Active/Pro-Caspase 3 Antibody in Western Blot (WB)
          Active/Pro-Caspase 3 Antibody in Western Blot (WB)
          Active/Pro-Caspase 3 Antibody in Western Blot (WB)
          Active/Pro-Caspase 3 Antibody in Western Blot (WB)
          Active/Pro-Caspase 3 Antibody in Immunohistochemistry (IHC)
          Active/Pro-Caspase 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Active/Pro-Caspase 3 Antibody in Immunohistochemistry (IHC)
          Active/Pro-Caspase 3 Antibody in Western Blot (WB)
          Active/Pro-Caspase 3 Antibody in Western Blot (WB)
          Active/Pro-Caspase 3 Antibody in Western Blot (WB)
          Active/Pro-Caspase 3 Antibody in Western Blot (WB)
          Active/Pro-Caspase 3 Antibody in Western Blot (WB)
          Active/Pro-Caspase 3 Antibody
          Active/Pro-Caspase 3 Antibody
          Active/Pro-Caspase 3 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Active/Pro-Caspase 3 Monoclonal Antibody (CPP32 4-1-18)

          Product Details

          MA1-16843

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 6 publications 6 publications

          Immunohistochemistry (IHC)

          -
          View 9 publications 9 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:10-1:500
          View 3 publications 3 publications

          Immunocytochemistry (ICC/IF)

          2 µg/mL
          -

          Immunoprecipitation (IP)

          2 µg/mg lysate
          -

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Insect, Mouse, Rabbit, Rat

          Host/Isotype

          Mouse / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          CPP32 4-1-18

          Immunogen

          Full-length recombinant human Caspase 3.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.0 mg/mL

          Amount

          200 µg

          Purification

          Protein G

          Storage buffer

          PBS

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_568479

          Product Specific Information

          Suggested positive control: antigen standard for CASP3 (transient overexpression lysate), HEK 293 whole cell lysate.

          Target Information

          This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. This protein cleaves and activates caspases 6, 7 and 9, and the protein itself is processed by caspases 8, 9 and 10. It is the predominant caspase involved in the cleavage of amyloid-beta 4A precursor protein, which is associated with neuronal death in Alzheimer's disease. Alternative splicing of this gene results in two transcript variants that encode the same protein.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Activated Caspase 3; Active Casp3; apopain; CASP-3; caspase 3, apoptosis related cysteine protease; Caspase 3, apoptosis related cysteine protease (ICE-like cysteine protease); caspase 3, apoptosis-related cysteine peptidase; caspase 3, apoptosis-related cysteine protease; Cleaved Caspase-3; Cleaved-Caspase3; CPP-32; CPP32beta; cysteine protease CPP32; cysteine protease p32-beta; IRP; PARP cleavage protease; procaspase3; protein Yama; SCA-1; SREBP cleavage activity 1; unnamed protein product

          View more View less

          Gene Aliases: A830040C14Rik; AC-3; CASP-3; Caspase-3; CC3; CPP-32; CPP32; CPP32-beta; CPP32B; Lice; mldy; SCA-1; Yama

          View more View less

          UniProt ID: (Human) P42574, (Mouse) P70677, (Rat) P55213

          View more View less

          Entrez Gene ID: (Human) 836, (Mouse) 12367, (Rat) 25402

          View more View less

          Function(s)
          protease binding endopeptidase activity aspartic-type endopeptidase activity cysteine-type endopeptidase activity cyclin-dependent protein serine/threonine kinase inhibitor activity death receptor binding protein binding enzyme activator activity peptidase activity cysteine-type peptidase activity phospholipase A2 activator activity macromolecular complex binding
          Process(es)
          luteolysis response to hypoxia negative regulation of cytokine production proteolysis apoptotic process axonal fasciculation learning or memory intrinsic apoptotic signaling pathway in response to osmotic stress response to xenobiotic stimulus response to glucose response to metal ion response to X-ray regulation of macroautophagy protein processing hippocampus development protein catabolic process neuron differentiation keratinocyte differentiation erythrocyte differentiation platelet formation regulation of protein stability response to cobalt ion response to estradiol response to lipopolysaccharide regulation of protein localization glial cell apoptotic process response to tumor necrosis factor response to nicotine intracellular signal transduction swimming behavior response to hydrogen peroxide positive regulation of apoptotic process response to amino acid regulation of neuron apoptotic process positive regulation of neuron apoptotic process response to ethanol neurotrophin TRK receptor signaling pathway striated muscle cell differentiation response to glucocorticoid protein maturation pyroptosis leukocyte apoptotic process response to anesthetic cellular response to staurosporine apoptotic signaling pathway intrinsic apoptotic signaling pathway execution phase of apoptosis regulation of synaptic vesicle cycle synapse pruning positive regulation of pyroptotic inflammatory response positive regulation of beta-amyloid formation positive regulation of apoptotic DNA fragmentation response to insulin-like growth factor stimulus B cell homeostasis cellular response to DNA damage stimulus heart development sensory perception of sound response to UV response to wounding negative regulation of B cell proliferation interleukin-18-mediated signaling pathway T cell homeostasis negative regulation of apoptotic process cell fate commitment negative regulation of cyclin-dependent protein serine/threonine kinase activity negative regulation of cell cycle negative regulation of activated T cell proliferation neuron apoptotic process anterior neural tube closure intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress protein poly-ADP-ribosylation response to ionizing radiation response to auditory stimulus animal organ development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7bf6cff7cf-hhljg:80/100.66.77.101:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0