Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 14
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
MA1-2010 detects the neurofilament, light chain in human, rat, and mouse samples.
MA1-2010 has been successfully used in western blot, immunohistochemistry, and immunofluorescence. By western blot this antibody detects a ~70 kDa protein representing the neurofilament, light chain in mouse brain tissue lysate. In immunohistochemistry procedures MA1-2010 recognizes the neurofilament, light chain in mouse brain tissue. In immunofluorescence procedures MA1-2010 recognizes the neurofilament, light chain in rat cerebral cortices.
The MA1-2010 immunogen is enzymatically dephosphorylated porcine neurofilaments.
Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-L is the light or low molecular weight microfilament subunit and runs on SDS-PAGE gels at approximately 70 kDa. Neurofilament are the 10nm or intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. NF-H is the heavy or high molecular weight microfilament subunit and runs on SDS-PAGE gels in the range 180-220 kDa, with some variation in different species.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 68 kDa neurofilament protein; 68kDa neurofilament; FLJ53642; intermediate filament; light molecular weight neurofilament protein; N-FL; NEF3; NEFL; neurofilament; neurofilament 3; neurofilament protein; neurofilament protein, light chain; neurofilament subunit NF-L; Neurofilament triplet L protein; neurofilament, light polypeptide; neurofilament, light polypeptide 68kDa; neurofilament-l; neurofilament-light; NF-M; NFM; NLC; OTTHUMP00000196614; protein phosphatase 1, regulatory subunit 110; unnamed protein product
Gene Aliases: CMT1F; CMT2E; CMTDIG; NF-L; NF68; NFL; PPP1R110
UniProt ID: (Human) P07196, (Rat) P19527, (Mouse) P08551
Entrez Gene ID: (Human) 4747, (Pig) 100521224, (Rat) 83613, (Mouse) 18039
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support