Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • NF-H Antibodies

          Invitrogen

          NF-H Monoclonal Antibody (3G3)

          Advanced Verification
          11 References
          View all (58) NF-H antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite NF-H Monoclonal Antibody (3G3)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          • Advanced Verification (1)
          NF-H Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NF-H Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NF-H Antibody (MA1-2012) in ICC/IF

          Immunofluorescence of neurofilament, heavy chain in rat cerebral cortex cultures in green. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NF-H Antibody in Immunocytochemistry (ICC/IF)
          NF-H Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NF-H Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NF-H Antibody in Western Blot (WB)
          NF-H Antibody in Western Blot (WB)
          NF-H Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          NF-H Monoclonal Antibody (3G3)

          Product Details

          MA1-2012

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:100-1:1,000
          -

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:10-1:100
          -

          Immunocytochemistry (ICC/IF)

          5 µg/mL
          -

          Immunoprecipitation (IP)

          -
          View 10 publications 10 publications
          Product Specifications

          Species Reactivity

          Human, Rat

          Published species

          Fruit fly, Human, Mouse, Rabbit, Rat

          Host/Isotype

          Mouse

          Class

          Monoclonal

          Type

          Antibody

          Clone

          3G3

          Immunogen

          Synthetic peptide corresponding to 226 amino acids of the C-terminus of neurofilament, heavy chain.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          purified

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_347005

          Product Specific Information

          MA1-2012 detects the neurofilament, heavy chain in human and rat samples.

          MA1-2012 has been successfully used in Western blot, immunohistochemistry and immunofluorecence procedures. By Western blot this antibody detects a ~200 kDa protein representing the neurofilament, heavy chain in rat brain microsome. In immunohistochemistry procedures MA1-2012 recognizes the neurofilament, heavy chain in rat hippocampal neurons.

          The MA1-2012 immunogen is a synthetic peptide corresponding to 226 amino acids of the C-terminus of neurofilament, heavy chain.

          Target Information

          Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-H has an important role in mature axons and is involved in amyotrophic lateral sclerosis (ALS).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 200 kDa neurofilament protein; heavy neurofilament polypeptide; heavy neurofilament subunit; high molecular weight neurofilament; hSTEp; NEF3; neurofilament; neurofilament triplet H protein; neurofilament, heavy polypeptide; neurofilament, heavy polypeptide 200kDa; NF 200 kD; NF-H; NHC; unnamed protein product

          View more View less

          Gene Aliases: CMT2CC; NFH

          View more View less

          UniProt ID: (Human) P12036

          View more View less

          Entrez Gene ID: (Human) 4744, (Rat) 24587

          View more View less

          Function(s)
          structural constituent of cytoskeleton protein binding microtubule binding kinesin binding protein kinase binding protein binding, bridging dynein complex binding structural constituent of postsynaptic intermediate filament cytoskeleton toxic substance binding macromolecular complex binding
          Process(es)
          axonogenesis cell projection assembly neurofilament bundle assembly intermediate filament bundle assembly axon development postsynaptic modulation of chemical synaptic transmission postsynaptic intermediate filament cytoskeleton organization regulation of organelle transport along microtubule microtubule cytoskeleton organization ovarian follicle atresia spinal cord development hippocampus development cerebral cortex development axon regeneration neuron projection development cellular response to oxidative stress response to cocaine intermediate filament cytoskeleton organization peripheral nervous system neuron axonogenesis neurofilament cytoskeleton organization cellular response to estradiol stimulus cellular response to hydroperoxide response to sodium arsenite response to acrylamide cellular response to leukemia inhibitory factor
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nv6df:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline