Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • 6x His Tag Antibodies

          Invitrogen

          6x-His Tag Monoclonal Antibody (HIS.H8)

          Advanced Verification
          72 Published Figures
          328 References
          View all (137) 6x His Tag antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite 6x-His Tag Monoclonal Antibody (HIS.H8)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (8)
          • Published Figures (72)
          • Advanced Verification (1)
          6x-His Tag Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          6x-His Tag Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 81

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          6x-His Tag Antibody (MA1-21315-1MG) in ICC/IF

          LEFT: untransfected control; RIGHT: anti-His (in red) on His-tagged fusion proteins in HEK293 cells. Both counterstained with DAPI (in blue) {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          6x-His Tag Antibody in Immunocytochemistry (ICC/IF)
          6x-His Tag Antibody in Immunocytochemistry (ICC/IF)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Immunocytochemistry (ICC/IF)
          6x-His Tag Antibody in Immunocytochemistry (ICC/IF)
          6x-His Tag Antibody in Immunocytochemistry (ICC/IF)
          6x-His Tag Antibody in Immunocytochemistry (ICC/IF)
          6x-His Tag Antibody in Immunocytochemistry (ICC/IF)
          6x-His Tag Antibody in Immunocytochemistry (ICC/IF)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Western Blot (WB)
          6x-His Tag Antibody in Immunoprecipitation (IP)
          6x-His Tag Antibody in Immunoprecipitation (IP)
          6x-His Tag Antibody in Immunoprecipitation (IP)
          6x-His Tag Antibody in ELISA (ELISA)
          6x-His Tag Antibody in ELISA (ELISA)
          6x-His Tag Antibody in ELISA (ELISA)
          6x-His Tag Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA1-21315-1MG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:3,000
          View 221 publications 221 publications

          Immunohistochemistry (IHC)

          -
          View 6 publications 6 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          1:500-1:2,000
          View 18 publications 18 publications

          Flow Cytometry (Flow)

          -
          View 9 publications 9 publications

          ELISA (ELISA)

          Assay-dependent
          View 35 publications 35 publications

          Immunoprecipitation (IP)

          Assay-dependent
          View 23 publications 23 publications

          Neutralization (Neu)

          -
          View 1 publication 1 publication

          Functional Assay (Functional)

          -
          View 2 publications 2 publications

          Peptide Array (Array)

          -
          View 2 publications 2 publications

          Inhibition Assays (Inhibition)

          -
          View 1 publication 1 publication

          GST Pull Down (GST-PD)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 7 publications 7 publications
          Product Specifications

          Species Reactivity

          Tag

          Published species

          Tag

          Host/Isotype

          Mouse / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          HIS.H8

          Immunogen

          6x His synthetic peptide.

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 555 
          • Alexa Fluor 647 
          • Biotin 
          • DyLight 488
          • DyLight 550
          • DyLight 650
          • DyLight 680
          • HRP
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          1 mg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.2

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2536982

          Product Specific Information

          MA1-21315 may be stored at 4° C short term. Add 0.05% sodium azide if desired. For long term storage, store at -20° C, avoiding freeze/thaw cycles.

          Target Information

          The 6x His tag is a synthetic oligo peptide consisting of 6 consecutive histidine residues (HHHHHH). The His tag is commonly expressed as a tag at either N-or C-terminal regions of recombinant proteins to allow isolation or purification by immobilized metal affinity chromatography. Epitope tagging is a technique in which a known epitope is fused to a recombinant protein using genetic engineering. By choosing a particular epitope and recombinant protein combination, epitope tagging makes it possible to detect proteins for which no antibody is available. His epitope tagged proteins that contain a stretch of histidine residues at the carboxyl or amino terminus are used for purification of recombinant proteins by means of metal chelate chromatography due to metal binding properties of histidine clusters on His tagged proteins. For example, the affinity of the histidine-tag motif to Ni2+ by chelation is strong and selective enough to enable purification of the protein to homogeneity by affinity chromatography on a Ni2+-NTA adsorbent. His tag-specific antibodies are used to facilitate detection or coimmunopreciptation of tagged proteins.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 6His tag; 6X His; His Tag; Histidine Tag

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch