Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 19
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Positive control of HELA WCE suggested.
Hec1 is one of several proteins involved in spindle checkpoint signaling. This surveillance mechanism assures correct segregation of chromosomes during cell division by detecting unaligned chromosomes and causing prometaphase arrest until the proper bipolar attachment of chromosomes is achieved.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HEC-1; HEC2; HEC4; highly expressed in cancer protein; highly expressed in cancer, rich in leucine heptad repeats; kinetochore associated 2; kinetochore protein Hec1; kinetochore protein NDC80 homolog; kinetochore-associated protein 2; NDC80 homolog, kinetochore complex component; NDC80 kinetochore complex component; NDC80 kinetochore complex component homolog; nuclear protein; putative kinetochore protein NDC80 like protein; retinoblastoma-associated protein HEC; unnamed protein product
Gene Aliases: 2610020P18Rik; HEC; HEC1; HsHec1; hsNDC80; KNTC2; TID3
UniProt ID: (Human) O14777, (Mouse) Q9D0F1
Entrez Gene ID: (Human) 10403, (Mouse) 67052, (Chinese hamster) 100751668
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support