Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Actin Antibodies

          Invitrogen

          Actin Monoclonal Antibody (AC-40), FITC

          View all (22) Actin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Actin Monoclonal Antibody (AC-40), FITC

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          Actin Monoclonal Antibody (AC-40), FITC

          Product Details

          MA1-24908

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          1:75
          -
          Product Specifications

          Species Reactivity

          Hamster, Human, Mollusc, Mouse, Sheep, Pig, Rabbit, Reptile, Rat, Xenopus, Bovine, Dog, Chicken, Fish, Guinea pig, Goat

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          AC-40

          Immunogen

          Synthetic actin C-terminal peptide Ser-Gly-Pro-Ser-Ile-Val-His-Arg-Lys-Cys-Phe, attached to a Multiple Antigen Peptide (MAP) backbone.
          3D Epitope / Immunogen

          Conjugate

          FITC FITC FITC
          • Cyanine3
          • Request custom conjugation

          Excitation/Emission Max

          498/517 nm View spectra spectra

          Form

          Liquid

          Concentration

          2.9 mg/mL

          Amount

          145 µg

          Purification

          purified

          Storage buffer

          PBS, pH 7.4, with 1% BSA

          Contains

          15mM sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_779142

          Product Specific Information

          Recommended positive controls: MDCK, human or chicken fibroblast.

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          Actin exists as a ubiquitous protein involved with filament formation that make up large portions of the cytoskeleton. Actin filaments interact with myosin to assist in muscle contraction as well as aiding in cell motility and cytokinesis. In vertebrates there are three groups of actin isoforms: alpha, beta and gamma. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. The beta and gamma actins co-exists in most cell types as components of the cytoskeleton and as mediators of internal cell motility.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: a-actin; ACT2; ACTA1; ACTA2; ACTB; ACTC; ACTG2; actin; actin 2; actin 7; actin alpha 1; actin alpha cardiac; actin alpha cardiac 1; actin homolog; Actin is a protein with a high degree of evolutionary conservation that undergoes polymerization to generate filaments, which subsequently assemble into interconnected networks within the cytoplasm of cells.; actin, alpha 1, skeletal muscle; actin, alpha 2, smooth muscle, aorta; Actin, alpha cardiac muscle 1; Actin, alpha skeletal muscle; actin, alpha, cardiac 1; actin, alpha, cardiac muscle 1; actin, alpha, vascular smooth muscle; Actin, aortic smooth muscle; actin, beta, cytoplasmic; Actin, cytoplasmic 1; Actin, cytoplasmic 2; Actin, cytoplasmic type 5; actin, cytoplasmic, type 5; actin, gamma 1; actin, gamma 1 propeptide; actin, gamma 2, smooth muscle, enteric; actin, gamma, cytoplasmic 1; Actin, gamma-enteric smooth muscle; actin-like protein; alpha 1 skeletal muscle actin; alpha actin; alpha actin 1; alpha actin 2; Alpha-actin; alpha-actin cardiac; Alpha-actin-1; Alpha-actin-2; Alpha-actin-3; Alpha-cardiac actin; alpha-sma; alpha-smooth muscle actin; alphac-actin; AtACT7; Atcb; B-actin; Bact; actin; beta actin; beta cytoskeletal actin; Beta-actin; beta-actin FE-3; cardiac actin; cardiac muscle alpha actin 1; Cell growth-inhibiting gene 46 protein; cytoplasmic 1; cytoplasmic 2; cytoplasmic actin; Cytoplasmic beta-actin; cytoplasmic type 5; cytoskeletal beta actin; cytoskeletal gamma-actin; cytoskeletal protein; DEFORMED ROOT HAIRS 1; DER1; ENHANCER OF LRX1 2; ENL2; epididymis luminal protein 176; FIZ2; FRIZZY AND KINKED SHOOTS 2; gamma 1 propeptide; gamma non-muscle actin; Gamma-2-actin; Gamma-actin; gamma-enteric smooth muscle actin; GIG46; I(2)-actin; LIGHT STRESS-REGULATED 2; LSR2; MYH9.2; MYH9_2; nemaline myopathy type 3; pan actin; PS1TP5-binding protein 1; RP5-1068B5.2; skeletal alpha actin; smooth muscle alpha-actin; Smooth muscle gamma-actin; type 5; unnamed protein product; vascular smooth muscle alpha-actin

          View more View less

          Gene Aliases: 0610041G09Rik; a-SMA; AA959943; ACT; ACT-4; ACT-5; Act4; ACTA; Acta-2; ACTA1; ACTA2; ACTA3; ACTB; ACTC; Actc-1; ACTC1; ACTE; ACTG; ACTG1; ACTG2; ACTGE; actin; Actl; ACTL3; Acts; ACTSA; ACTSG; Actsk-1; ACTVS; Actx; AL023024; alphaSMA; ASD5; ASMA; Bact; Beta-actin; BKRNS; BNS; BRWS1; CFTD; CFTD1; CFTDM; CMD1R; CMH11; CMYO2A; CMYO2B; CMYO2C; CMYP2A; CMYP2B; CMYP2C; CSMH; DDS1; DFNA20; DFNA26; E430023M04Rik; E51; GAMMA-ACTIN; GIG46; HEL-176; I79_002310; I79_013242; I79_019066; LVNC4; MMIHS5; MPFD; NEM1; NEM2; NEM3; PS1TP5BP1; SHPM; SMalphaA; SMDYS; SMGA; THC8; VSCM; VSCM1; XELAEV_18029136mg; XELAEV_18045052mg

          View more View less

          UniProt ID: (Rabbit) P68135, (Human) P68133, (Chicken) P68139, (Bovine) P68138, (Pig) P68137, (Bovine) P62739, (Human) P62736, (Rabbit) P62740, (Chicken) P08023, (Pig) Q6QAQ1, (Human) P60709, (Chicken) P60706, (Dog) O18840, (Rabbit) P29751, (Bovine) P60712, (Sheep) P60713, (Chicken) P68034, (Human) P68032, (Bovine) Q3ZC07, (Chicken) P53478, (Bovine) P63258, (Human) P63261, (Bovine) Q5E9B5, (Human) P63267, (Chicken) P63270, (Mouse) P68134, (Rat) P68136, (Mouse) P62737, (Rat) P62738, (Rat) P60711, (Guinea pig) Q71FK5, (Mouse) P60710, (Chinese hamster) P48975, (Rat) P68035, (Mouse) P68033, (Rat) P63259, (Mouse) P63260, (Mouse) P63268, (Rat) P63269, (Xenopus) O93400

          View more View less

          Entrez Gene ID: (Rabbit) 100009506, (Sheep) 101107662, (Goat) 102182476, (Human) 58, (Chicken) 421534, (Bovine) 281592, (Pig) 100154254, (Dog) 488984, (Pig) 733615, (Goat) 102191486, (Bovine) 515610, (Human) 59, (Dog) 477587, (Rabbit) 100009271, (Chicken) 423787, (Sheep) 101121051, (Pig) 414396, (Human) 60, (Chicken) 396526, (Dog) 403580, (Rabbit) 100009272, (Bovine) 280979, (Sheep) 443052, (Goat) 102179831, (Dog) 478250, (Chicken) 423298, (Sheep) 101111554, (Human) 70, (Pig) 100152267, (Bovine) 533219, (Goat) 102170606, (Rabbit) 100340339, (Goat) 100860883, (Chicken) 415296, (Bovine) 404122, (Pig) 397653, (Sheep) 443340, (Dog) 475923, (Human) 71, (Goat) 102178279, (Bovine) 281595, (Human) 72, (Dog) 475792, (Sheep) 101114256, (Chicken) 396084, (Pig) 100520667, (Rabbit) 100350318, (Chinese hamster) 100768972, (Mouse) 11459, (Guinea pig) 100716933, (Rat) 29437, (Mouse) 11475, (Rat) 81633, (Chinese hamster) 100758575, (Guinea pig) 100722917, (Rat) 81822, (Guinea pig) 100135470, (Mouse) 11461, (Chinese hamster) 100689477, (Rat) 29275, (Guinea pig) 100731953, (Mouse) 11464, (Chinese hamster) 100766388, (Chinese hamster) 100764022, (Rat) 287876, (Mouse) 11465, (Guinea pig) 100731713, (Mouse) 11468, (Chinese hamster) 100768744, (Guinea pig) 100716255, (Rat) 25365, (Xenopus) 432314, (Xenopus) 398459

          View more View less

          Function(s)
          nucleotide binding structural constituent of cytoskeleton protein binding ATP binding hydrolase activity myosin binding ADP binding protein kinase binding ATPase activity kinesin binding nitric-oxide synthase regulator activity Tat protein binding nucleosomal DNA binding identical protein binding tau protein binding nitric-oxide synthase binding structural constituent of postsynaptic actin cytoskeleton transporter regulator activity microfilament motor activity profilin binding ubiquitin protein ligase binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II distal enhancer sequence-specific DNA binding molecular_function actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein
          Process(es)
          muscle contraction positive regulation of gene expression skeletal muscle thin filament assembly skeletal muscle fiber development mesenchyme migration regulation of blood pressure response to virus vascular smooth muscle contraction glomerular mesangial cell development positive regulation of hepatic stellate cell migration positive regulation of hepatic stellate cell contraction positive regulation of ERK1 and ERK2 cascade cellular response to transforming growth factor beta stimulus juxtaglomerular apparatus development positive regulation of hepatic stellate cell activation postsynaptic actin cytoskeleton organization morphogenesis of a polarized epithelium chromatin remodeling regulation of transcription from RNA polymerase II promoter cytoskeleton organization establishment or maintenance of cell polarity axonogenesis positive regulation of cell proliferation substantia nigra development regulation of mitotic metaphase/anaphase transition adherens junction assembly maintenance of blood-brain barrier regulation of apoptotic process apical protein localization positive regulation of T cell differentiation negative regulation of cell differentiation positive regulation of cell differentiation positive regulation of myoblast differentiation positive regulation of transcription, DNA-templated cell motility regulation of norepinephrine uptake positive regulation of norepinephrine uptake regulation of cell cycle regulation of G0 to G1 transition platelet aggregation protein localization to adherens junction cellular response to cytochalasin B regulation of transepithelial transport regulation of synaptic vesicle endocytosis positive regulation of stem cell population maintenance regulation of protein localization to plasma membrane positive regulation of double-strand break repair via homologous recombination regulation of G1/S transition of mitotic cell cycle regulation of double-strand break repair positive regulation of double-strand break repair regulation of nucleotide-excision repair regulation of transmembrane transporter activity negative regulation of protein binding synaptic vesicle endocytosis actin filament organization actin-myosin filament sliding heart contraction response to xenobiotic stimulus actin filament-based movement actomyosin structure organization negative regulation of apoptotic process response to ethanol cardiac myofibril assembly cardiac muscle tissue morphogenesis cardiac muscle contraction cytoplasmic actin-based contraction involved in cell motility angiogenesis positive regulation of cell migration regulation of stress fiber assembly regulation of focal adhesion assembly positive regulation of wound healing tight junction assembly protein localization to bicellular tight junction sarcomere organization cellular response to interferon-gamma response to mechanical stimulus response to extracellular stimulus response to lithium ion cell growth skeletal muscle fiber adaptation response to steroid hormone retina homeostasis ATP-dependent chromatin remodeling retina development in camera-type eye cellular response to electrical stimulus metabolic process response to drug actin-mediated cell contraction response to calcium ion biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline