Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: mouse whole brain.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta-a gap junction protein; connexin 32; connexin g; connexin-32; connexin-43; GAP junction 28 kDa liver protein; gap junction membrane channel protein beta 1; gap junction protein, beta 1, 32kDa; unnamed protein product
Gene Aliases: CMTX; CMTX1; Cnx32; connexin-32; CX32; Cxng
UniProt ID: (Human) P08034, (Mouse) P28230, (Rat) P08033
Entrez Gene ID: (Human) 2705, (Mouse) 14618, (Rat) 29584
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support