Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 8
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody reacts with an antigen present in cutaneous melanocytes, prenatal and infantile retinal pigment epithelium and melanoma cells.This antibody reacts with melanomas and tumors showing melanoma/melanocytic differentiation. It reacts with junctional and blue nevus cells. Non-melanocytic cells do not react with this antibody.
Melanoma is a malignant tumor of melanocytes which are found predominantly in skin but also in the bowel and the eye (see uveal melanoma). It is one of the rarer types of skin cancer but causes the majority of skin cancer related deaths.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 95kDa melanocyte-specific secreted glycoprotein; Melanocyte lineage specific antigen GP100; melanocyte lineage-specific antigen/Pmel17 homolog; melanocyte protein mel 17; melanocyte protein Pmel 17; melanocyte-specific secreted glycoprotein; melanocytes lineage-specific antigen GP100; melanoma-associated ME20 antigen; melanosomal matrix protein17; Pmel 17; silver locus protein homolog; silver, mouse, homolog of; unnamed protein product
Gene Aliases: D12S53E; gp100; HMB-45; HMB45; ME20; ME20-M; ME20M; P1; P100; PMEL17; SI; SIL; SILV
UniProt ID: (Human) P40967
Entrez Gene ID: (Human) 6490
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support