Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody is specific for the ~55 kDa THR-beta protein.
Thyroid hormone receptors (TR), also known as c-erbA-2, are ligand-dependent, intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences called hormone response elements following activation by the appropriate hormone. Thyroid hormones, through their interaction with TR, effect metabolic processes, growth and development in many tissues by regulating the expression of genes for growth hormone, malic enzyme and several hepatic proteins. There are two forms of TR; TR alpha (on chromosome 17) and TR beta (on chromosome 3). Each of these isoforms also has two isoforms; TR alpha-1 and TRv alpha-2, and TR beta-1 and TR beta-2 respectively. TR alpha-1 and 2 are identical through amino acid 370 where their sequences diverge. TRv alpha-2, which does not bind T3 and is a strong negative regulator of the functional TR, is 80 amino acids longer than TR alpha-1. The corresponding gene for the thyroid hormone receptor beta-1 is NR1A2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ERBA 2; ERBA BETA; ERBA-BETA; MGC126109; MGC126110; nuclear receptor; nuclear receptor subfamily 1 group A member 2; oncogene ERBA2; thyroid hormone receptor, beta (erythroblastic leukemia viral (v-erb-a) oncogene homolog 2, avian); TR; TR beta 1; TR-B1C-ERBA-2; unnamed protein product
Gene Aliases: C-ERBA-2; C-ERBA-BETA; ERBA2; GRTH; NR1A2; PRTH; THR1; THRB1; THRB2; THRbeta; THRbeta1; Thrbeta2; TRb; TRbeta; TRbeta1
UniProt ID: (Human) P10828
Entrez Gene ID: (Human) 7068
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support