Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 53
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
MA1-710 detects vitamin D receptor (VDR) from human, rat, mouse, hamster, chicken, amphibian, and fish tissues. This antibody does not cross-react with estrogen or glucocorticoid receptors. MA1-710 detects both the occupied and unoccupied forms of the receptor.
MA1-710 has been successfully used in Western blot, IF, ICC, IP, ChIP, and gel shift procedures. By Western blot, this antibody detects a 58 kDa and 60 kDa protein representing VDR in avians while the VDRs of mammalian species appear as single bands within the 52-55 kDa range.
The MA1-710 immunogen is partially purified chicken intestinal cytoplasmic 1,25-(OH)2D3 receptor protein. This antibody recognizes an epitope between amino acid residues 89 and 105 of human VDR. This region is just C-terminal to the DNA binding domain of the VDR.
Steroid receptors are ligand-dependent intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate hormone. The 1,25-dihydroxy-vitamin D3 receptor (VDR) belongs to the superfamily of steroid hormone receptors which includes estrogen, progesterone, glucocorticoid, androgen, and thyroid hormone receptors. Vitamin D modulates calcium and phosphorus homeostasis, bone remodeling, cell growth regulation, and differentiation. Studies have found VDR in the intestine, bone, kidney, epidermis, and cells of the endocrine immune system. Repression of T-cell proliferation and interleukin-2 (IL-2) gene expression, for example, occurs when VDR binds within the IL-2 enhancer. Formation of VDR/retinoic X receptor (RXR) heterodimers in the presence of intracellular 1,25(OH)2D3 has been shown to interfere with assembly of nuclear factor of activated T-cells (NFATp)/Fos/Jun/DNA complex and subsequent IL-2 gene transcription. The corresponding gene for the vitamin D receptor is NR1I1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1,25-dihydroxyvitamin D-3 receptor; 1,25-dihydroxyvitamin D3 receptor; HGNC:12679; nuclear receptor subfamily 1 group I member 1; protein phosphatase 1, regulatory subunit 163; unnamed protein product; Vdr; vitamin D (1,25- dihydroxyvitamin D3) receptor; vitamin D receptor
Gene Aliases: NR1I1; PPP1R163
UniProt ID: (Chicken) O42392, (Human) P11473, (Mouse) P48281, (Rat) P13053
Entrez Gene ID: (Chicken) 395988, (Human) 7421, (Mouse) 22337, (Rat) 24873
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support