Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
MA1-7620 detects Glutathione from all organism samples.
MA1-7620 has been successfully used in ELISA, WB and IP procedures.
The MA1-7620 immunogen is a Glutathione protein.
Store at 4°C short term. For long term storage, store at -80°C
Glutathione is a tripeptide comprised of three amino acids (cysteine, glutamic acid, and glycine) present in most mammalian tissue. Glutathione acts as an antioxidant, a free radical scavenger and a detoxifying agent. Glutathione is also important as a cofactor for the enzyme glutathione peroxidase, in the uptake of amino acids, and in the synthesis of leukotrienes. As a substrate for glutathione S-transferase, this agent reacts with a number of harmful chemical species, such as halides, epoxides and free radicals, to form harmless inactive products. In erythrocytes, these reactions prevent oxidative damage through the reduction of methemoglobin and peroxides. Glutathione is also involved in the formation and maintenance of disulfide bonds in proteins and in the transport of amino acids across cell membranes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C10H17N3O6S; Glutathion; Glutathione-SH; GPX4; L-Glutathione; Reduced glutathione
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support