Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 5
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody does not react with dog.
This product requires enzyme-mediated antigen retrieval prior to staining of paraffin sections. Proteinase K is recommended.
Mouse anti glucose transporter 4 antibody, clone 1F8 originally raised against rat intracellular low density microsomes (James et al.
This gene is a member of the solute carrier family 2 (facilitated glucose transporter) family and encodes a protein that functions as an insulin-regulated facilitative glucose transporter. In the absence of insulin, this integral membrane protein is sequestered within the cells of muscle and adipose tissue. Within minutes of insulin stimulation, the protein moves to the cell surface and begins to transport glucose across the cell membrane. Mutations in this gene have been associated with noninsulin-dependent diabetes mellitus (NIDDM).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glucose transporter 2; glucose transporter 4; glucose transporter 4 insulin-responsive; Glucose transporter 4, insuline-responsive; glucose transporter glut4; glucose transporter type 4; glucose transporter type 4, insulin-responsive; GLUT-4; insulin responsive glucose transporter; insulin-responsive glucose transporter; insulin-responsive glucose transporter type 4; MGC93607; solute carrier family 2 (facilitated glucose transporter), member 4; solute carrier family 2 , member 4; unnamed protein product
Gene Aliases: Glut-4; GLUT4; GT2; twgy
UniProt ID: (Human) P14672, (Mouse) P14142, (Rat) P19357
Entrez Gene ID: (Rabbit) 100037710, (Human) 6517, (Pig) 396754, (Mouse) 20528, (Rat) 25139
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support