Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PMCA4 ATPase Antibodies

          Invitrogen

          PMCA4 ATPase Monoclonal Antibody (JA9)

          Advanced Verification
          11 Published Figures
          19 References
          View all (6) PMCA4 ATPase antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PMCA4 ATPase Monoclonal Antibody (JA9)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          • Published Figures (11)
          • Advanced Verification (2)
          PMCA4 ATPase Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PMCA4 ATPase Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 16

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PMCA4 ATPase Antibody (MA1-914) in ICC/IF

          Immunolocalization of PMCA4 and CaSRO in bovine corneal epithelium (bCE). (A) Immunolocalization of PMCA4 (green) using Product # MA1-914 on cell membranes of all cells in each layer of bCE. (B) Immunolocalization of CaSRO (red) using Product # MA1-934 on cell membranes of bCE mostly in wing- and squamous cell layers. (C) Merged image. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PMCA4 ATPase Antibody in Immunocytochemistry (ICC/IF)
          PMCA4 ATPase Antibody in Immunocytochemistry (ICC/IF)
          PMCA4 ATPase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PMCA4 ATPase Antibody in Immunocytochemistry (ICC/IF)
          PMCA4 ATPase Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          PMCA4 ATPase Antibody in Immunocytochemistry (ICC/IF)
          PMCA4 ATPase Antibody in Immunohistochemistry (IHC)
          PMCA4 ATPase Antibody in Immunohistochemistry (IHC)
          PMCA4 ATPase Antibody in Western Blot (WB)
          PMCA4 ATPase Antibody in Western Blot (WB)
          PMCA4 ATPase Antibody in Western Blot (WB)
          PMCA4 ATPase Antibody in Western Blot (WB)
          PMCA4 ATPase Antibody in Western Blot (WB)
          PMCA4 ATPase Antibody in Western Blot (WB)
          PMCA4 ATPase Antibody
          PMCA4 ATPase Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          PMCA4 ATPase Monoclonal Antibody (JA9)

          Product Details

          MA1-914

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 12 publications 12 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:500
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 4 publications 4 publications

          Flow Cytometry (Flow)

          2 µg / 10^6 cells
          -

          ELISA (ELISA)

          1:15,000
          -
          Product Specifications

          Species Reactivity

          Bovine, Human, Pig, Rat

          Published species

          Amphibian, Bovine, Human, Mouse, Pig, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          JA9

          Immunogen

          Purified human erythrocyte PMCA.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          purified

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2061596

          Product Specific Information

          MA1-914 specifically detects plasma membrane calcium pump PMCA4 ATPase from human, bovine, porcine and rat tissues.

          MA1-914 has been successfully used in Western blot, immunohistochemistry and ELISA procedures. By Western blot, this antibody detects an ~129 kDa and an ~133 kDa protein representing PMCA4b and PMCA4a ATPase, respectively.

          The MA1-914 antigen is purified human erythrocyte PMCA ATPase. The epitope for this antibody has been mapped to amino acids 51-75 of PMCA4 ATPase. This sequence is common to both PMCA4a and 4b ATPase.

          Target Information

          The calcium pump of the plasma membrane, termed PMCA ATPase, pumps calcium from the cytosol to the extracellular space. This membrane-bound enzyme is related to a number of other ATPases including the SERCA ATPase and the sodium/K+ pump. There are four different genes encoding PMCA ATPase and studies have revealed 20 isoforms of the pump generated by alternate splicing of the primary gene products. mRNA distribution studies show that gene products 1 and 4 are transcribed in most tissues, however, products 2 and 3 are more tissue specific. Transcription of the splicing variants has also been found to be tissue specific. In the pancreas, where insulin secretion is calcium dependent, the beta cells only express the 4b isoform, however the alpha and gamma cells express both 4a and 4b isoforms. Studies have also shown that different splice variants have different affinities for calcium and calmodulin (Cat. # MA3-917 and MA3-918).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ATPase, Ca++ transporting, plasma membrane 4; ATPase, Ca++ transporting, plasma membrane 4 ATP synthase, H+ transporting, mitochondrial F1; ATPase, Ca++ transporting, plasma membrane 4 tv2; matrix-remodeling-associated protein 1; plasma membrane Ca2+-ATPases 4; plasma membrane calcium ATPase; plasma membrane calcium ATPase 4; plasma membrane calcium pump; plasma membrane calcium-ATPase isoform 4 alternative splice variant x at splice site A; plasma membrane calcium-ATPase isoform 4, splice variant b at alternative splice site C; plasma membrane calcium-pumping ATP-ase; PMCA4b; sarcolemmal calcium pump; unnamed protein product

          View more View less

          Gene Aliases: ATP2B2; MXRA1; PMCA4; PMCA4b; PMCA4x

          View more View less

          UniProt ID: (Human) P23634, (Bovine) D3K0R6, (Rat) Q64542

          View more View less

          Entrez Gene ID: (Human) 493, (Bovine) 282146, (Pig) 100623420, (Rat) 29600

          View more View less

          Function(s)
          nucleotide binding calcium-transporting ATPase activity protein binding calmodulin binding ATP binding calcium ion transmembrane transporter activity ATPase activity sodium channel regulator activity protein kinase binding PDZ domain binding protein phosphatase 2B binding nitric-oxide synthase inhibitor activity nitric-oxide synthase binding scaffold protein binding metal ion binding calcium-dependent protein binding primary active transporter
          Process(es)
          neural retina development regulation of transcription from RNA polymerase II promoter calcium ion transport cellular calcium ion homeostasis spermatogenesis negative regulation of gene expression urinary bladder smooth muscle contraction negative regulation of angiogenesis hippocampus development sperm motility ion transmembrane transport nitric oxide-cGMP-mediated signaling pathway negative regulation of blood vessel endothelial cell migration negative regulation of nitric oxide biosynthetic process regulation of cytosolic calcium ion concentration response to hydrostatic pressure calcium ion transmembrane transport negative regulation of calcineurin-NFAT signaling cascade cellular response to epinephrine stimulus negative regulation of adrenergic receptor signaling pathway calcium ion transmembrane import into cytosol calcium ion import across plasma membrane negative regulation of the force of heart contraction transport across blood-brain barrier negative regulation of arginine catabolic process calcium ion export regulation of sodium ion transmembrane transport negative regulation of cellular response to vascular endothelial growth factor stimulus regulation of cell cycle G1/S phase transition positive regulation of protein localization to plasma membrane negative regulation of cardiac muscle hypertrophy in response to stress negative regulation of citrulline biosynthetic process regulation of cardiac conduction cellular response to acetylcholine negative regulation of nitric oxide mediated signal transduction positive regulation of peptidyl-serine phosphorylation negative regulation of nitric-oxide synthase activity negative regulation of adenylate cyclase-activating adrenergic receptor signaling pathway involved in heart process positive regulation of cAMP-dependent protein kinase activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch