Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • DYKDDDDK Tag Antibodies

          Invitrogen

          DYKDDDDK Tag Monoclonal Antibody (FG4R)

          Advanced Verification
          61 Published Figures
          174 References
          View all (127) DYKDDDDK Tag antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite DYKDDDDK Tag Monoclonal Antibody (FG4R)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          • Published Figures (61)
          • Advanced Verification (1)
          DYKDDDDK Tag Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          DYKDDDDK Tag Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 66

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DYKDDDDK Tag Antibody (MA1-91878) in ICC/IF

          Immunofluorescent analysis of Chlamydia trachomatis FLAG®-tagged IncA (blue) in HeLa cells. Cells were infected with a GFP-expressing strain of Chlamydia trachomatis containing FLAG-IncA under the control of the Tetracycline (Tet) promoter, and either left untreated (0 ng/mL Tet) or treated with anhydrotetracycline (10 ng/mL Tet). Cells were fixed with 2% paraformaldehyde in PBS for 30 minutes at room tempe... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DYKDDDDK Tag Antibody in Immunocytochemistry (ICC/IF)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Immunoprecipitation (IP)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Immunocytochemistry (ICC/IF)
          DYKDDDDK Tag Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          DYKDDDDK Tag Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          DYKDDDDK Tag Antibody in Immunocytochemistry (ICC/IF)
          DYKDDDDK Tag Antibody in Immunocytochemistry (ICC/IF)
          DYKDDDDK Tag Antibody in Immunocytochemistry (ICC/IF)
          DYKDDDDK Tag Antibody in Western Blot, Immunocytochemistry, Immunoprecipitation (WB, ICC/IF, IP)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          DYKDDDDK Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          DYKDDDDK Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          DYKDDDDK Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot, ELISA (WB, ELISA)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Western Blot (WB)
          DYKDDDDK Tag Antibody in Immunoprecipitation (IP)
          DYKDDDDK Tag Antibody in ELISA (ELISA)
          DYKDDDDK Tag Antibody in ELISA (ELISA)
          DYKDDDDK Tag Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA1-91878

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 109 publications 109 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:200-1:500
          View 26 publications 26 publications

          Flow Cytometry (Flow)

          -
          View 2 publications 2 publications

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          3 µg
          View 27 publications 27 publications

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication

          Peptide Array (Array)

          -
          View 1 publication 1 publication

          Gel Shift (EMSA)

          -
          View 1 publication 1 publication

          RNA Immunoprecipitation (RIP)

          -
          View 1 publication 1 publication

          In Situ Hybridization (ISH)

          -
          View 1 publication 1 publication

          CUT&RUN (CUT&RUN)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Tag

          Published species

          Tag

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          FG4R

          Immunogen

          Synthetic peptide (DYKDDDDK) coupled to KLH.

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Biotin 
          • DyLight 488
          • DyLight 550
          • DyLight 650
          • DyLight 680
          • DyLight 800 4X PEG
          • HRP
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.2

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1957945

          Product Specific Information

          This antibody detects Flag tag from various samples.

          similar epitope as the FLAG tag from Sigma

          MA1-91878 has been successfully used in immunoprecipitation, Western blot and immunofluorescence applications.

          The MA1-91878 immunogen is synthetic peptide (DYKDDDDK) coupled to KLH.

          MA1-91878 may be stored at 4°C short term. Add 0.05% sodium azide if desired. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Target Information

          DYKDDDDK tag, also known as FLAG tag, is a short peptide sequence that is commonly used as a protein tag to facilitate protein purification, detection, and localization. The tag consists of eight amino acids: Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys. It is often fused to the N- or C-terminus of a protein of interest, allowing for easy identification and isolation of the protein using anti-FLAG antibodies. The DYKDDDDK tag is widely used in molecular biology and biochemistry research, and has been employed in various applications, such as protein-protein interaction studies, protein expression analysis, and protein localization studies. The FLAG tag structure has been optimized for compatibility with the proteins it is attached to, in that it is more hydrophilic than other common epitope tags and therefore less likely to denature or inactivate proteins to which it is appended. FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: DDDDK; DDDDK epitope tag; ECS epitope tag; ECS tag; FLAG; FLAG Tag

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch