Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Ryanodine Receptor Antibodies

          Invitrogen

          Ryanodine Receptor Monoclonal Antibody (C3-33)

          Advanced Verification
          45 Published Figures
          251 References
          View all (6) Ryanodine Receptor antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Ryanodine Receptor Monoclonal Antibody (C3-33)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Published Figures (45)
          • Advanced Verification (1)
          Ryanodine Receptor Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          Ryanodine Receptor Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 48

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Ryanodine Receptor Antibody (MA3-916) in IHC

          Immunofluorescent analysis of rat brain hippocampus tissue shows that Ryanodine Receptor Monoclonal Antibody (Product # MA3-916) crossreacts with RyR1. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Ryanodine Receptor Antibody in Immunohistochemistry (IHC)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Immunocytochemistry (ICC/IF)
          Ryanodine Receptor Antibody in Immunocytochemistry (ICC/IF)
          Ryanodine Receptor Antibody in Immunocytochemistry (ICC/IF)
          Ryanodine Receptor Antibody in Immunocytochemistry (ICC/IF)
          Ryanodine Receptor Antibody in Immunocytochemistry (ICC/IF)
          Ryanodine Receptor Antibody in Immunocytochemistry (ICC/IF)
          Ryanodine Receptor Antibody in Immunohistochemistry (IHC)
          Ryanodine Receptor Antibody in Immunohistochemistry (IHC)
          Ryanodine Receptor Antibody in Immunohistochemistry (IHC)
          Ryanodine Receptor Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Ryanodine Receptor Antibody in Immunohistochemistry (IHC)
          Ryanodine Receptor Antibody in Immunohistochemistry (IHC)
          Ryanodine Receptor Antibody in Immunohistochemistry (IHC)
          Ryanodine Receptor Antibody in Immunohistochemistry (IHC)
          Ryanodine Receptor Antibody in Immunohistochemistry (IHC)
          Ryanodine Receptor Antibody in Immunohistochemistry (IHC)
          Ryanodine Receptor Antibody in Immunohistochemistry (IHC)
          Ryanodine Receptor Antibody in Immunohistochemistry (IHC)
          Ryanodine Receptor Antibody in Immunohistochemistry (IHC)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Western Blot (WB)
          Ryanodine Receptor Antibody in Immunoprecipitation (IP)
          Ryanodine Receptor Antibody in Immunoprecipitation (IP)
          Ryanodine Receptor Antibody

          Product Details

          MA3-916

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          View 137 publications 137 publications

          Immunohistochemistry (IHC)

          -
          View 42 publications 42 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1 µg/mL
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          1 µg/mL
          View 4 publications 4 publications

          Immunocytochemistry (ICC/IF)

          1:50
          View 53 publications 53 publications

          Flow Cytometry (Flow)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          View 10 publications 10 publications

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          Dot blot (DB)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Amphibian, Dog, Chicken, Fish, Guinea pig, Human, Mouse, Rabbit, Rat

          Published species

          Dog, Guinea pig, Hamster, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          C3-33

          Immunogen

          Canine cardiac ryanodine receptor (RyR2)

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          PBS

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2183054

          Product Specific Information

          MA3-916 detects ryanodine receptor 2 (RyR2) and shows cross-reactivity with RyR1 in amphibian, canine, chicken, fish, guinea pig and rat tissues.

          MA3-916 has been successfully used in Western blot, immunohistochemical, immunofluorescence, immunocytochemistry and immunoprecipitation procedures. By Western blot, this antibody detects a 565 kDa protein representing the ryanodine receptor. In non-mammalian vertebrates, a doublet is seen at 565 kDa representing the alpha and beta isoforms of the receptor. Immunohistochemical staining of RyR in rat cardiac tissue with MA3-916 results in intense staining of the myofiber, which is consistent with sarcoplasmic reticulum localization.

          Target Information

          The ryanodine receptor (RyR) is the channel responsible for the release of calcium from the sarcoplasmic reticulum (SR) in muscle cells and also plays a role in calcium regulation in non-muscle cells. The RyR exists as a homotetramer and is predicted to have a short cytoplasmic C-terminus and 4-10 transmembrane domains; the remainder of the protein, termed the "foot" region is located in the cytoplasm between the T-tubule and the SR. The mammalian RyR is the product of three different genes: RyR-1, which is expressed predominantly in skeletal muscle and areas of the brain, RyR-2, which is expressed predominantly in the heart muscle but also found in the stomach, endothelial cells and diffuse areas of the brain, and RyR-3 which is found in smooth muscle and the brain (striatum, thalamus and hippocampus).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Ca2+ release channel; calcium release channel; Cardiac muscle ryanodine receptor; Cardiac muscle ryanodine receptor-calcium release channel; cardiac muscle-type ryanodine receptor; cardiac ryanodine receptor; cardiac ryanodine receptor 2; cardiac-type ryanodine receptor; central core disease of muscle; islet-type ryanodine receptor; kidney-type ryanodine receptor; protein phosphatase 1, regulatory subunit 137; ryanodine receptor; Ryanodine receptor 1; ryanodine receptor 1 (skeletal); ryanodine receptor 1, skeletal muscle; ryanodine receptor 1-like, skeletal muscle; Ryanodine receptor 2; ryanodine receptor 2 (cardiac); ryanodine receptor type 2; ryanodine receptor type II; RYR-1; RYR-2; sarcoplasmic reticulum calcium release channel; Skeletal muscle calcium release channel; Skeletal muscle ryanodine receptor; Skeletal muscle-type ryanodine receptor; Type 1 ryanodine receptor; type 1-like ryanodine receptor; Type 2 ryanodine receptor; unnamed protein product

          View more View less

          Gene Aliases: 9330127I20Rik; AI528790; ARVC2; ARVD2; CCO; CMYO1A; CMYO1B; CMYP1A; CMYP1B; KDS; MHS; MHS1; PPP1R137; RYDR; RYR; RYR-1; RYR-2; RYR1; Ryr1l; RYR2; SKRR; VACRDS; VTSIP

          View more View less

          UniProt ID: (Rabbit) P11716, (Human) P21817, (Human) Q92736, (Mouse) E9PZQ0, (Rat) B0LPN4, (Mouse) E9Q401

          View more View less

          Entrez Gene ID: (Rabbit) 100009540, (Human) 6261, (Dog) 606491, (Rabbit) 100009536, (Chicken) 421508, (Human) 6262, (Dog) 403615, (Mouse) 20190, (Guinea pig) 100727848, (Rat) 114207, (Rat) 689560, (Mouse) 20191

          View more View less

          Function(s)
          nucleotide binding ion channel activity ryanodine-sensitive calcium-release channel activity voltage-gated calcium channel activity calcium channel activity calcium ion binding calmodulin binding ATP binding calcium-release channel activity metal ion binding calcium-induced calcium release activity protein binding enzyme binding protein kinase binding protein kinase A catalytic subunit binding protein kinase A regulatory subunit binding identical protein binding suramin binding ion channel binding protease binding protein self-association scaffold protein binding organic cyclic compound binding ligand-gated ion channel scaffold/adaptor protein
          Process(es)
          response to hypoxia outflow tract morphogenesis ion transport calcium ion transport cellular calcium ion homeostasis muscle contraction striated muscle contraction release of sequestered calcium ion into cytosol by sarcoplasmic reticulum calcium-mediated signaling response to caffeine ion transmembrane transport skin development ossification involved in bone maturation skeletal muscle fiber development release of sequestered calcium ion into cytosol protein homotetramerization regulation of cytosolic calcium ion concentration transmembrane transport calcium ion transmembrane transport cellular response to calcium ion cellular response to caffeine regulation of heart rate embryonic heart tube morphogenesis left ventricular cardiac muscle tissue morphogenesis cardiac muscle hypertrophy detection of calcium ion positive regulation of heart rate regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion regulation of cardiac muscle contraction by calcium ion signaling response to muscle activity response to muscle stretch positive regulation of sequestering of calcium ion establishment of localization in cell response to redox state regulation of cardiac muscle contraction cardiac muscle contraction calcium ion transport into cytosol sarcoplasmic reticulum calcium ion transport cellular response to epinephrine stimulus establishment of protein localization to endoplasmic reticulum ventricular cardiac muscle cell action potential Purkinje myocyte to ventricular cardiac muscle cell signaling cell communication by electrical coupling involved in cardiac conduction type B pancreatic cell apoptotic process calcium ion transmembrane import into cytosol positive regulation of the force of heart contraction regulation of AV node cell action potential regulation of SA node cell action potential regulation of atrial cardiac muscle cell action potential regulation of ventricular cardiac muscle cell action potential transport regulation of muscle contraction multicellular organism development regulation of ion transmembrane transport response to nutrient BMP signaling pathway response to magnesium ion calcium-mediated signaling using intracellular calcium source response to drug negative regulation of cytosolic calcium ion concentration response to calcium ion canonical Wnt signaling pathway cytosolic calcium ion transport manganese ion transmembrane transport positive regulation of ryanodine-sensitive calcium-release channel activity by adrenergic receptor signaling pathway involved in positive regulation of cardiac muscle contraction positive regulation of calcium-transporting ATPase activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-7c59b9b899-7wltc:80/100.66.72.81:80.
          git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.49.0-Offline