Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SERCA1 ATPase Antibodies

          Invitrogen

          SERCA1 ATPase Monoclonal Antibody (IIH11)

          20 Published Figures
          80 References
          View all (19) SERCA1 ATPase antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SERCA1 ATPase Monoclonal Antibody (IIH11)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          • Published Figures (20)
          SERCA1 ATPase Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SERCA1 ATPase Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 26

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SERCA1 ATPase Antibody (MA3-911) in ICC/IF

          Immunofluorescent analysis of SERCA1 ATPase using Anti-SERCA1 ATPase Monoclonal Antibody (IIH11) (Product # MA3-911) shows staining in C6 Cells. SERCA1 ATPase staining (green), F-Actin staining with Phalloidin (red) and nuclei with DAPI (blue) is shown. Cells were grown on chamber slides and fixed with formaldehyde prior to staining. Cells were probed without (control) or with or an antibody recognizing SER... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SERCA1 ATPase Antibody in Immunocytochemistry (ICC/IF)
          SERCA1 ATPase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SERCA1 ATPase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SERCA1 ATPase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SERCA1 ATPase Antibody in Immunohistochemistry (IHC)
          SERCA1 ATPase Antibody in Western Blot (WB)
          SERCA1 ATPase Antibody in Immunohistochemistry (IHC)
          SERCA1 ATPase Antibody in Immunohistochemistry (IHC)
          SERCA1 ATPase Antibody in Immunohistochemistry (IHC)
          SERCA1 ATPase Antibody in Western Blot (WB)
          SERCA1 ATPase Antibody in Western Blot (WB)
          SERCA1 ATPase Antibody in Western Blot (WB)
          SERCA1 ATPase Antibody in Western Blot (WB)
          SERCA1 ATPase Antibody in Western Blot (WB)
          SERCA1 ATPase Antibody in Western Blot (WB)
          SERCA1 ATPase Antibody in Western Blot (WB)
          SERCA1 ATPase Antibody in Western Blot (WB)
          SERCA1 ATPase Antibody in Western Blot (WB)
          SERCA1 ATPase Antibody in Western Blot (WB)
          SERCA1 ATPase Antibody in Western Blot (WB)
          SERCA1 ATPase Antibody in Western Blot (WB)
          SERCA1 ATPase Antibody in Western Blot (WB)
          SERCA1 ATPase Antibody in Western Blot (WB)
          SERCA1 ATPase Antibody in Western Blot (WB)
          SERCA1 ATPase Antibody in Western Blot (WB)
          SERCA1 ATPase Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          SERCA1 ATPase Monoclonal Antibody (IIH11)

          Product Details

          MA3-911

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:5,000
          View 62 publications 62 publications

          Immunohistochemistry (IHC)

          -
          View 16 publications 16 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:500
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Dog, Guinea pig, Human, Mouse, Rabbit, Rat

          Published species

          Dog, Human, Invertebrate, Mouse, Rabbit, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          IIH11

          Immunogen

          Purified rabbit skeletal muscle sarcoplasmic reticulum.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          PBS, ascites

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325494

          Product Specific Information

          MA3-911 detects sarcoplasmic or endoplasmic reticulum calcium 1 (SERCA1) ATPase from canine, human, rabbit, rat, mouse and guinea pig tissues.

          MA3-911 has been successfully used in Western blot, immunohistochemistry, and immunofluorescence procedures. By Western blot, this antibody detects an ~110 kDa protein representing SERCA1 ATPase in canine skeletal muscle extracts. Immunofluorescence staining of SERCA1 ATPase in canine skeletal muscle with MA3-911 results in strong labeling of the entire type II (fast) myofiber.

          The MA3-911 antigen is purified rabbit skeletal muscle sarcoplasmic reticulum. This antibody recognizes an epitope between amino acids 199-505 of rabbit skeletal muscle ATPase, a region that is not exposed in native sarcoplasmic reticulum.

          Target Information

          ATP dependent calcium pumps are responsible, in part, for the maintenance of low cytoplasmic free calcium concentrations. The ATP pumps that reside in intracellular organelles are encoded by a family of structurally related enzymes, termed the sarcoplasmic or endoplasmic reticulum calcium (SERCA) ATPases. The SERCA1 gene is exclusively expressed in type II (fast) skeletal muscle. The SERCA2 gene is subject to tissue dependent processing which is responsible for the generation of SERCA2a muscle-specific form expressed in type I (slow) skeletal, cardiac and smooth muscle and the SERCA2b isoform expressed in all cell types. The SERCA3 gene is not as well characterized and is found in non-muscle cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: adult Ca2+ ATPase; ATPase, Ca++ transporting, cardiac muscle, fast twitch 1; ATPase, Ca++ transporting, ubiquitous; Ca2+ ATPase; calcium pump 1; endoplasmic reticulum class 1/2 Ca(2+) ATPase; neonatal Ca2+ ATPase; sarco/endoplasmic reticulum calcium ATPase 1; sarco/endoplasmic reticulum calcium ATPase 1a; sarco/endoplasmic reticulum calcium ATPase 1b; SERCA; SERCA ATPase; SERCA1; SR Ca(2+)-ATPase 1; unnamed protein product

          View more View less

          Gene Aliases: ATP2A; ATP2A3; SERCA1; SERCA1a

          View more View less

          UniProt ID: (Human) O14983, (Rabbit) P04191, (Mouse) Q8R429, (Rat) Q64578

          View more View less

          Entrez Gene ID: (Dog) 479797, (Human) 487, (Rabbit) 100037716, (Mouse) 11937, (Rat) 116601, (Guinea pig) 100726234

          View more View less

          Function(s)
          nucleotide binding calcium-transporting ATPase activity calcium ion binding protein binding ATP binding ATPase activity calcium-dependent ATPase activity protein homodimerization activity nucleotide-sugar transmembrane transporter activity primary active transporter
          Process(es)
          calcium ion transport cellular calcium ion homeostasis regulation of striated muscle contraction apoptotic mitochondrial changes positive regulation of fast-twitch skeletal muscle fiber contraction positive regulation of endoplasmic reticulum calcium ion concentration negative regulation of endoplasmic reticulum calcium ion concentration ion transmembrane transport response to endoplasmic reticulum stress negative regulation of striated muscle contraction positive regulation of mitochondrial calcium ion concentration maintenance of mitochondrion location intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress calcium ion import calcium ion transmembrane transport relaxation of skeletal muscle positive regulation of cardiac muscle cell contraction positive regulation of calcium-transporting ATPase activity positive regulation of calcium ion import into sarcoplasmic reticulum regulation of cardiac conduction calcium ion import into sarcoplasmic reticulum regulation of muscle contraction organophosphate ester transport nucleobase-containing compound transport response to peptide hormone sarcoplasmic reticulum calcium ion transport carbohydrate derivative transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-nnlvt:80/100.66.74.151:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0