Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Calpain 1 Antibodies

          Invitrogen

          Mu-Calpain Monoclonal Antibody (9A4H8D3)

          Advanced Verification
          9 Published Figures
          52 References
          View all (26) Calpain 1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Mu-Calpain Monoclonal Antibody (9A4H8D3)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (7)
          • Published Figures (9)
          • Advanced Verification (1)
          Mu-Calpain Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Mu-Calpain Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 17

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Mu-Calpain Antibody (MA3-940) in ICC/IF

          Immunofluorescent analysis of Mu-Calpain in MCF-7 Cells. Cells were grown on chamber slides and fixed with formaldehyde prior to staining. Cells were probed without (control) or with a Mu-Calpain monoclonal antibody (Product # MA3-940) at a dilution of 1:20 overnight at 4 C, washed with PBS and incubated with a DyLight-488 conjugated secondary antibody (Product # 35503). Mu-Calpain staining (green), F-Actin... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Mu-Calpain Antibody in Immunocytochemistry (ICC/IF)
          Mu-Calpain Antibody in Immunocytochemistry (ICC/IF)
          Mu-Calpain Antibody in Immunocytochemistry (ICC/IF)
          Mu-Calpain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Mu-Calpain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Mu-Calpain Antibody in Western Blot (WB)
          Mu-Calpain Antibody in Western Blot (WB)
          Mu-Calpain Antibody in Immunocytochemistry (ICC/IF)
          Mu-Calpain Antibody in Western Blot (WB)
          Mu-Calpain Antibody in Western Blot (WB)
          Mu-Calpain Antibody in Western Blot (WB)
          Mu-Calpain Antibody in Western Blot (WB)
          Mu-Calpain Antibody in Western Blot (WB)
          Mu-Calpain Antibody in Western Blot (WB)
          Mu-Calpain Antibody in Western Blot (WB)
          Mu-Calpain Antibody in Western Blot (WB)
          Mu-Calpain Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Mu-Calpain Monoclonal Antibody (9A4H8D3)

          Product Details

          MA3-940

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          2 µg/mL
          View 47 publications 47 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-Dependent
          -

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Bovine, Hamster, Human, Mouse, Pig, Rabbit, Rat

          Published species

          Arthropod, Bovine, Fish, Hamster, Human, Mouse, Pig, Rabbit, Rat, Sheep, Yeast

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          9A4H8D3

          Immunogen

          Purified bovine skeletal muscle 80 kDa mu-calpain subunit.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.0 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2069338

          Product Specific Information

          MA3-940 detects mu-calpain from human platelets and erythrocytes, bovine platelets, heart and skeletal muscle, rat myoblasts, kidney, liver and spleen, mouse lung, pig cultured cells and hamster and rabbit samples. This antibody does not cross-react with m-calpain, n-calpain, calmodulin or calpastatin.

          MA3-940 has been successfully used in Western blot, immunofluorescence, immunohistochemistry, and immunocytochemistry procedures. By Western blot, this antibody detects an 80 kDa protein representing mu-calpain from human platelets and erythrocytes and HeLa and A431 cell lysates. Immunocytochemical staining of mu-calpain in LLC-PK1 cells with MA3-940 results in diffuse cytoplasmic staining. This product has not been shown to be effective in immunoprecipitation experiments.

          The MA3-940 antigen is purified bovine skeletal muscle 80 kDa mu-calpain subunit. This antibody recognizes an epitope between amino acids 465-520 (domain III) of human mu-calpain.

          Target Information

          The calpains including Calpain 1 (CAPN1), calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. Several transcript variants encoding two different isoforms have been found for this gene. Diseases associated with CAPN1 include Spastic Paraplegia 76, Autosomal Recessive and Spasticity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: calcium activated neutral protease; calcium-activated neutral protease 1; calcium-activated neutral proteinase 1; calcium-dependent protease; calpain 1 large subunit; calpain 1, (mu/I) large subunit; calpain 1, large subunit; calpain mu-type; calpain, large polypeptide L1; Calpain-1 catalytic subunit; calpain-1 large subunit; Calpain-I; Calpain-II; CANP 1; cell proliferation-inducing gene 30 protein; cell proliferation-inducing protein 30; micromolar calcium-activated neutral protease 1; micromolar-calpain; mu-calpain; mucalpain; muCANP; PIG30; protease; unnamed protein product

          View more View less

          Gene Aliases: CANP; CANP 1; CANP1; CANPL1; Capa-1; Capa1; I79_024883; mu-calpin; muCANP; muCL; SPG76

          View more View less

          UniProt ID: (Pig) P35750, (Bovine) Q27970, (Human) P07384, (Mouse) O35350, (Rat) P97571

          View more View less

          Entrez Gene ID: (Rabbit) 100009089, (Pig) 397027, (Bovine) 281661, (Human) 823, (Chinese hamster) 100758636, (Mouse) 12333, (Rat) 29153

          View more View less

          Function(s)
          calcium-dependent cysteine-type endopeptidase activity calcium ion binding protein binding peptidase activity cytoskeletal protein binding enzyme binding cysteine protease
          Process(es)
          proteolysis receptor catabolic process regulation of catalytic activity mammary gland involution self proteolysis positive regulation of cell proliferation regulation of macroautophagy NMDA selective glutamate receptor signaling pathway positive regulation of cardiac muscle cell apoptotic process protein autoprocessing protein catabolic process negative regulation of actin filament polymerization positive regulation of vascular permeability cellular response to hydrogen peroxide protein catabolic process at postsynapse negative regulation of NIK/NF-kappaB signaling positive regulation of leukocyte tethering or rolling response to angiotensin response to arsenic-containing substance
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7bf6cff7cf-x9r6b:80/100.66.78.146:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0