Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • TGF beta-1,2,3 Antibodies

          Invitrogen

          TGF beta-1,2,3 Monoclonal Antibody (1D11)

          Advanced Verification
          8 Published Figures
          11 References
          View all (12) TGF beta-1,2,3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TGF beta-1,2,3 Monoclonal Antibody (1D11)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Published Figures (8)
          • Advanced Verification (1)
          TGF beta-1,2,3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TGF beta-1,2,3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 9

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TGF beta-1,2,3 Antibody (MA5-23795) in ICC/IF

          Cancers 2021 - Figure 6 Effect of TGF-beta1 neutralizing antibody and latrunculin B in the Boyden chamber and 3D sphere assay. ( A ) Immunofluorescence images of the Boyden chamber assay. ( B , C ) Images of representative immunofluorescence results revealed that the addition of TGF-beta1-blocking antibody or latrunculin B reduced actin stress fibers and vimentin intermediate filament networks. BFTC909 cells were seeded i... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TGF beta-1,2,3 Antibody in Immunocytochemistry (ICC/IF)
          TGF beta-1,2,3 Antibody in Neutralization (Neu)
          TGF beta-1,2,3 Antibody in Neutralization (Neu)
          TGF beta-1,2,3 Antibody in Neutralization (Neu)
          TGF beta-1,2,3 Antibody in Neutralization (Neu)
          TGF beta-1,2,3 Antibody in Neutralization (Neu)
          TGF beta-1,2,3 Antibody in Neutralization (Neu)
          TGF beta-1,2,3 Antibody in Inhibition Assays (Inhibition)
          TGF beta-1,2,3 Antibody
          TGF beta-1,2,3 Monoclonal Antibody (1D11)

          Product Details

          MA5-23795

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 1 publication 1 publication

          ELISA (ELISA)

          0.1-0.4 µg/mL (Detection), 2-8 µg/mL (Capture)
          -

          Neutralization (Neu)

          0.25-1.25 µg/mL
          View 4 publications 4 publications

          Functional Assay (Functional)

          -
          View 1 publication 1 publication

          Inhibition Assays (Inhibition)

          Assay-Dependent
          View 3 publications 3 publications

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Bovine, Chicken, Human, Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1D11

          Immunogen

          Bovine bone-derived TGF-beta 1 and TGF-beta 2
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.5 mg/mL

          Amount

          500 µg

          Purification

          Protein A/G

          Storage buffer

          PBS, pH 7.4, with 5% trehalose

          Contains

          No Preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2609812

          Product Specific Information

          This antibody recognizes human TGF-beta 1, TGF-beta 2, and TGF-beta 3.

          Reconstitute at 0.5 mg/mL in sterile PBS.

          Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.

          Neutralization: measured by its ability to neutralize TGF beta-1 inhibition of IL-4-dependent proliferation in the HT-2 mouse T cell line. The Neutralization Dose (ND50) is typically 0.25-1.25 µg/mL in the presence of 1 ng/mL Recombinant Human TGF beta-1 and 7.5 ng/mL Recombinant Mouse IL-4.

          Target Information

          TGF-beta1, -2, and -3 are a closely related group of proteins (70-80% sequence homology) that are produced by many cell types and function as growth and differentiation factors. The active forms of TGF-beta1, -2, and -3 are disulfide-linked homodimers.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: BSC-1 cell growth inhibitor; Cetermin; G-TSF; Glioblastoma-derived T-cell suppressor factor; latency-associated peptide; MGF; Milk growth factor; polyergin; prepro-transforming growth factor beta-1; prepro-transforming growth factor beta-2; prepro-transforming growth factor beta-3; regulatory protein; tgf beta 1; TGF beta 3; tgf beta1; TGF beta2; TGF-beta 1; TGF-beta-1; TGF-beta-2; TGF-beta-3; tissue growth factor-beta4; Transforming growth factor; transforming growth factor beta 4; transforming growth factor beta precursor; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor beta-2; Transforming growth factor beta-2 proprotein; transforming growth factor beta-3; Transforming growth factor beta-3 proprotein; transforming growth factor beta1; transforming growth factor, beta 1 (Camurati-Engelmann disease); transforming growth factor-beta 1; transforming growth factor-beta-1 precursor; transforming growth factor-beta-2 precursor; transforming growth factor-beta2; transforming growth factor-beta3; unnamed protein product

          View more View less

          Gene Aliases: ARVD; ARVD1; BB105277; CED; DPD1; G-TSF; IBDIMDE; LAP; LDS4; LDS5; MGF; RNHF; TGF-beta-2; TGF-beta-3; TGF-beta1; TGF-beta2; TGF-beta3; TGF-beta4; TGFB; Tgfb-1; Tgfb-2; Tgfb-3; TGFB1; TGFB2; TGFB3; TGFB4; TGFbeta; TGFbeta1

          View more View less

          UniProt ID: (Human) P01137, (Bovine) P18341, (Bovine) P21214, (Human) P61812, (Human) P10600, (Chicken) P16047, (Mouse) P04202, (Mouse) P27090

          View more View less

          Entrez Gene ID: (Human) 7040, (Bovine) 282089, (Chicken) 100873157, (Bovine) 534069, (Chicken) 421352, (Human) 7042, (Human) 7043, (Chicken) 396438, (Bovine) 538957, (Mouse) 21803, (Mouse) 21808, (Mouse) 21809

          View more View less

          Function(s)
          type II transforming growth factor beta receptor binding cytokine activity transforming growth factor beta receptor binding protein binding growth factor activity enzyme binding type I transforming growth factor beta receptor binding type III transforming growth factor beta receptor binding deubiquitinase activator activity identical protein binding protein serine/threonine kinase activator activity macromolecular complex binding antigen binding beta-amyloid binding receptor binding protein homodimerization activity transforming growth factor beta binding glycoprotein binding protein heterodimerization activity protein N-terminus binding receptor signaling protein serine/threonine kinase activity growth factor
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter cell morphogenesis vasculogenesis ureteric bud development response to hypoxia morphogenesis of a branching structure cell activation epithelial to mesenchymal transition neural tube closure regulation of sodium ion transport sprouting angiogenesis chondrocyte differentiation columnar/cuboidal epithelial cell maturation hematopoietic progenitor cell differentiation connective tissue replacement involved in inflammatory response wound healing adaptive immune response based on somatic recombination of immune receptors built from immunoglobulin superfamily domains tolerance induction to self antigen negative regulation of natural killer cell mediated cytotoxicity directed against tumor cell target heart valve morphogenesis aortic valve morphogenesis protein export from nucleus ATP biosynthetic process phosphate-containing compound metabolic process cellular calcium ion homeostasis apoptotic process inflammatory response transforming growth factor beta receptor signaling pathway Notch signaling pathway negative regulation of neuroblast proliferation salivary gland morphogenesis endoderm development heart development female pregnancy cell proliferation positive regulation of cell proliferation negative regulation of cell proliferation germ cell migration response to xenobiotic stimulus response to wounding response to glucose gene expression regulation of gene expression positive regulation of vascular endothelial growth factor production positive regulation of gene expression negative regulation of gene expression negative regulation of extracellular matrix disassembly positive regulation of epithelial to mesenchymal transition macrophage derived foam cell differentiation positive regulation of fibroblast migration negative regulation of macrophage cytokine production oligodendrocyte development positive regulation of microglia differentiation regulation of striated muscle tissue development neural tube development hyaluronan catabolic process T cell differentiation negative regulation of ossification negative regulation of cell growth osteoclast differentiation regulation of cell migration positive regulation of cell migration mammary gland development organ regeneration membrane protein intracellular domain proteolysis positive regulation of protein complex assembly positive regulation of exit from mitosis response to estradiol response to progesterone regulation of interleukin-23 production negative regulation of interleukin-17 production positive regulation of interleukin-17 production positive regulation of interleukin-6 production positive regulation of tumor necrosis factor production receptor catabolic process positive regulation of superoxide anion generation mononuclear cell proliferation regulation of actin cytoskeleton organization positive regulation of collagen biosynthetic process response to vitamin D response to laminar fluid shear stress response to immobilization stress myofibroblast differentiation T cell proliferation T cell activation regulation of cell proliferation negative regulation of T cell proliferation regulation of protein import into nucleus positive regulation of protein import into nucleus odontogenesis of dentin-containing tooth positive regulation of odontogenesis myelination T cell homeostasis positive regulation of apoptotic process positive regulation of vascular permeability positive regulation of I-kappaB kinase/NF-kappaB signaling surfactant homeostasis proteasome-mediated ubiquitin-dependent protein catabolic process positive regulation of MAPK cascade positive regulation of blood vessel endothelial cell migration negative regulation of blood vessel endothelial cell migration regulatory T cell differentiation cell-cell junction organization response to ethanol regulation of regulatory T cell differentiation positive regulation of regulatory T cell differentiation negative regulation of cell differentiation positive regulation of cell differentiation negative regulation of fat cell differentiation negative regulation of myoblast differentiation positive regulation of epidermal growth factor receptor signaling pathway negative regulation of cell cycle negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter muscle cell cellular homeostasis positive regulation of fibroblast proliferation lung alveolus development positive regulation of isotype switching to IgA isotypes lymph node development digestive tract development negative regulation of skeletal muscle tissue development positive regulation of smooth muscle cell proliferation inner ear development epithelial cell proliferation positive regulation of epithelial cell proliferation negative regulation of epithelial cell proliferation positive regulation of protein secretion positive regulation of inflammatory response negative regulation of phagocytosis defense response to fungus negative regulation of T cell activation positive regulation of chemotaxis positive regulation of cellular component organization positive regulation of smooth muscle cell differentiation positive regulation of multicellular organismal process positive regulation of protein metabolic process negative regulation of release of sequestered calcium ion into cytosol neuron apoptotic process regulation of cell cycle positive regulation of cell division positive regulation of protein kinase B signaling ventricular cardiac muscle tissue morphogenesis phospholipid homeostasis canonical Wnt signaling pathway regulation of blood vessel remodeling face morphogenesis frontal suture morphogenesis positive regulation of SMAD protein import into nucleus bronchiole development mammary gland branching involved in thelarche branch elongation involved in mammary gland duct branching regulation of branching involved in mammary gland duct morphogenesis regulation of cartilage development retina vasculature development in camera-type eye connective tissue development Langerhans cell differentiation negative regulation of biomineral tissue development regulation of enamel mineralization lens fiber cell differentiation positive regulation of ERK1 and ERK2 cascade response to cholesterol cellular response to mechanical stimulus cellular response to glucose stimulus cellular response to growth factor stimulus cellular response to low-density lipoprotein particle stimulus cellular response to hypoxia cellular response to ionizing radiation cellular response to dexamethasone stimulus cellular response to transforming growth factor beta stimulus positive regulation of mononuclear cell migration odontoblast differentiation stem cell proliferation extracellular matrix assembly positive regulation of branching involved in ureteric bud morphogenesis positive regulation of canonical Wnt signaling pathway extrinsic apoptotic signaling pathway liver regeneration cellular response to virus transforming growth factor beta receptor superfamily signaling pathway negative regulation of hyaluronan biosynthetic process positive regulation of protein localization to nucleus positive regulation of extracellular matrix assembly response to salt positive regulation of mesenchymal stem cell proliferation regulation of miRNA transcription negative regulation of pri-miRNA transcription from RNA polymerase II promoter positive regulation of pri-miRNA transcription from RNA polymerase II promoter negative regulation of protein localization to plasma membrane positive regulation of vasculature development positive regulation of STAT cascade regulation of epithelial to mesenchymal transition involved in endocardial cushion formation cellular response to acetaldehyde cellular response to insulin-like growth factor stimulus embryonic liver development negative regulation of cell-cell adhesion mediated by cadherin positive regulation of chemokine (C-X-C motif) ligand 2 production positive regulation of endothelial cell apoptotic process positive regulation of primary miRNA processing positive regulation of stem cell proliferation positive regulation of cardiac muscle cell differentiation MAPK cascade negative regulation of protein phosphorylation positive regulation of protein phosphorylation protein phosphorylation cell cycle arrest mitotic cell cycle checkpoint epidermal growth factor receptor signaling pathway common-partner SMAD protein phosphorylation SMAD protein complex assembly positive regulation of peptidyl-threonine phosphorylation positive regulation of pathway-restricted SMAD protein phosphorylation cell migration regulation of transforming growth factor beta receptor signaling pathway negative regulation of cell-cell adhesion positive regulation of bone mineralization lipopolysaccharide-mediated signaling pathway positive regulation of cellular protein metabolic process positive regulation of peptidyl-serine phosphorylation positive regulation of mammary gland epithelial cell proliferation positive regulation of protein dephosphorylation positive regulation of MAP kinase activity protein kinase B signaling positive regulation of phosphatidylinositol 3-kinase activity ossification involved in bone remodeling negative regulation of mitotic cell cycle regulation of binding regulation of DNA binding pathway-restricted SMAD protein phosphorylation regulation of SMAD protein import into nucleus cellular response to organic cyclic compound positive regulation of NAD+ ADP-ribosyltransferase activity positive regulation of transdifferentiation positive regulation of transcription regulatory region DNA binding BMP signaling pathway positive regulation of luteinizing hormone secretion positive regulation of phosphorylation progesterone secretion positive regulation of follicle-stimulating hormone secretion regulation of protein secretion SMAD protein signal transduction eye development hair follicle development heart morphogenesis cell death glial cell migration cardioblast differentiation positive regulation of epithelial cell migration negative regulation of alkaline phosphatase activity positive regulation of phosphatidylinositol 3-kinase signaling signal transduction by protein phosphorylation collagen fibril organization positive regulation of cell growth activation of protein kinase activity positive regulation of stress-activated MAPK cascade regulation of transforming growth factor beta2 production positive regulation of cell adhesion mediated by integrin response to drug positive regulation of neuron apoptotic process positive regulation of integrin biosynthetic process positive regulation of ossification positive regulation of heart contraction embryonic digestive tract development positive regulation of catagen positive regulation of cardioblast differentiation cardiac muscle cell proliferation cardiac epithelial to mesenchymal transition somitogenesis epithelial to mesenchymal transition involved in endocardial cushion formation negative regulation of cell adhesion nervous system development skeletal muscle tissue development gonad development male gonad development female gonad development response to virus embryo development ending in birth or egg hatching hemopoiesis lung development neutrophil chemotaxis regulation of growth wound healing endothelial cell activation dopamine biosynthetic process regulation of cell differentiation positive regulation of cell cycle somatic stem cell division animal organ development spleen development embryonic organ development neuron development positive regulation of immune response cartilage development response to growth hormone regulation of complement-dependent cytotoxicity skeletal system development cartilage condensation blood vessel development kidney development blood vessel remodeling outflow tract septum morphogenesis membranous septum morphogenesis outflow tract morphogenesis atrioventricular valve morphogenesis pulmonary valve morphogenesis endocardial cushion morphogenesis cardiac right ventricle morphogenesis ventricular trabecula myocardium morphogenesis endocardial cushion fusion atrial septum primum morphogenesis neural retina development activation-induced cell death of T cells axon guidance sensory organ development striated muscle tissue development negative regulation of angiogenesis signaling extracellular matrix organization embryonic limb morphogenesis epithelial cell differentiation hair follicle morphogenesis ascending aorta morphogenesis odontogenesis uterine wall breakdown regulation of apoptotic process positive regulation of Notch signaling pathway negative regulation of Ras protein signal transduction neuron fate commitment generation of neurons regulation of timing of catagen uterus development ventricular septum morphogenesis atrial septum morphogenesis negative regulation of cartilage development pharyngeal arch artery morphogenesis secondary palate development positive regulation of cardiac epithelial to mesenchymal transition positive regulation of activation-induced cell death of T cells positive regulation of extracellular matrix disassembly extrinsic apoptotic signaling pathway in absence of ligand regulation of apoptotic process involved in outflow tract morphogenesis regulation of extracellular matrix organization substantia propria of cornea development pericyte cell differentiation cranial skeletal system development negative regulation of epithelial to mesenchymal transition involved in endocardial cushion formation positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation positive regulation of extrinsic apoptotic signaling pathway in absence of ligand in utero embryonic development negative regulation of neuron apoptotic process positive regulation of filopodium assembly positive regulation of stress fiber assembly detection of hypoxia negative regulation of vascular smooth muscle cell proliferation positive regulation of occluding junction disassembly mesenchyme migration protein import into nucleus, translocation CD4-positive, CD25-positive, alpha-beta regulatory T cell lineage commitment defense response SMAD protein import into nucleus negative regulation of DNA replication defense response to fungus, incompatible interaction organ morphogenesis response to organic substance cell growth evasion or tolerance of host defenses by virus positive regulation of histone deacetylation regulation of CD4-positive, CD25-positive, alpha-beta regulatory T cell differentiation positive regulation of histone acetylation growth myeloid dendritic cell differentiation regulation of MAPK cascade cell development negative regulation of immune response positive regulation of NF-kappaB transcription factor activity negative regulation of gene silencing by miRNA positive regulation of cell cycle arrest T-helper 17 cell lineage commitment regulation of actin cytoskeleton reorganization cardiac left ventricle morphogenesis intracellular signal transduction catagen negative regulation of apoptotic process response to estrogen negative regulation of keratinocyte differentiation embryonic neurocranium morphogenesis activation of MAPK activity embryo development negative regulation of transforming growth factor beta receptor signaling pathway positive regulation of DNA replication palate development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline