Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
MA5-13395 targets Thrombospondin in FACS, IF, IHC (P), IM, IP, and WB applications and shows reactivity with Bovine, Canine, Equine, Human, mouse, Ovine, Porcine, and Rat samples.
The MA5-13395 immunogen is reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets.
Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glycoprotein G; Thrombospondin; thrombospondin-1p180; thrombospondin-p50; ThrombospondinTHBS1; TSP-1; unnamed protein product
Gene Aliases: tbsp1; THBS; THBS-1; TSP; TSP-1; TSP1
UniProt ID: (Human) P07996, (Bovine) Q28178
Entrez Gene ID: (Horse) 100057478, (Human) 7057, (Dog) 487486, (Bovine) 281530, (Pig) 492313, (Sheep) 101106433, (Rat) 445442, (Mouse) 21825
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support