Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PERK Antibodies

          Invitrogen

          Phospho-PERK (Thr980) Monoclonal Antibody (G.305.4)

          Advanced Verification
          17 Published Figures
          29 References
          View all (38) PERK antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-PERK (Thr980) Monoclonal Antibody (G.305.4)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          • Published Figures (17)
          • Advanced Verification (1)
          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 20

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-PERK (Thr980) Antibody (MA5-15033) in WB

          Western blot was performed using anti-Phospho-PERK (Thr980) Monoclonal Antibody (G.305.4) (Product # MA5-15033) and a 125 kDa band corresponding to Phospho-PERK was induced in RSC96 cells upon thapsigargin and tunicamycin treatments which are known to induce ER stress. Whole cell extracts (30 µg lysate) of RSC96 (Lane1), RSC96 treated with Thapsigargin (300nM for 2hrs) (Lane 2) and RSC96 with Tunicamycin (4... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Phospho-PERK (Thr980) Antibody in Immunohistochemistry (IHC)
          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Phospho-PERK (Thr980) Antibody in Western Blot (WB)
          Phospho-PERK (Thr980) Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-PERK (Thr980) Monoclonal Antibody (G.305.4)

          Product Details

          MA5-15033

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 23 publications 23 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Inhibition Assays (Inhibition)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Monoclonal

          Type

          Antibody

          Clone

          G.305.4

          Immunogen

          Synthetic phosphopeptide corresponding to residues surrounding pThr980 of mouse PERK
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          441 µg/mL

          Amount

          44.1 µg

          Purification

          Affinity chromatography

          Storage buffer

          0.01M HEPES, pH 7.5, with 0.15M NaCl, 100µg/mL BSA, 50% glycerol

          Contains

          <0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10980432

          Product Specific Information

          It is not recommended to aliquot this antibody.

          Target Information

          EIF2AK3 (PERK) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses, leading to a decrease in protein sythesis. In the case of EIF2AK3, signaling is initiated by misfolded proteins in the ER. Defects in EIF2AK3 are the cause of Wolcott-Rallison syndrome (WRS), also known as multiple epiphyseal dysplasia with early-onset diabetes mellitus. WRS is a rare autosomal recessive disorder, characterized by permanent neonatal or early infancy insulin-dependent diabetes and, at a later age, epiphyseal dysplasia, osteoporosis, growth retardation and other multisystem manifestations, such as hepatic and renal dysfunctions, mental retardation and cardiovascular abnormalities.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 2.7.11.1; eIF-2 alpha kinase; OTTHUMP00000207187; OTTHUMP00000207188; pancreatic eIF2-alpha kinase; PRKR-like endoplasmic reticulum kinase; protein tyrosine kinase EIF2AK3; unnamed protein product

          View more View less

          Gene Aliases: PEK; Perk

          View more View less

          UniProt ID: (Rat) Q9Z1Z1, (Mouse) Q9Z2B5

          View more View less

          Entrez Gene ID: (Rat) 29702, (Mouse) 13666

          View more View less

          Function(s)
          protein kinase activity protein serine/threonine kinase activity eukaryotic translation initiation factor 2alpha kinase activity protein tyrosine kinase activity non-membrane spanning protein tyrosine kinase activity ATP binding enzyme binding protein phosphatase binding identical protein binding translation regulator activity misfolded protein binding Hsp90 protein binding protein serine kinase activity protein binding eukaryotic initiation factor eIF2 binding
          Process(es)
          skeletal system development ossification angiogenesis chondrocyte development translation regulation of translational initiation protein phosphorylation ER overload response endoplasmic reticulum organization lactation positive regulation of signal transduction positive regulation of vascular endothelial growth factor production positive regulation of gene expression negative regulation of gene expression regulation of translational initiation by eIF2 alpha phosphorylation negative regulation of translation regulation of fatty acid metabolic process calcium-mediated signaling bone mineralization endoplasmic reticulum unfolded protein response pancreas development endocrine pancreas development negative regulation of myelination negative regulation of translation in response to stress SREBP signaling pathway cellular response to amino acid starvation response to endoplasmic reticulum stress regulation of translation initiation in response to endoplasmic reticulum stress eiF2alpha phosphorylation in response to endoplasmic reticulum stress PERK-mediated unfolded protein response ERAD pathway cellular response to glucose starvation negative regulation of apoptotic process regulation of translational initiation in response to stress fat cell differentiation positive regulation of transcription from RNA polymerase I promoter positive regulation of transcription from RNA polymerase II promoter protein autophosphorylation insulin-like growth factor receptor signaling pathway regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress cellular response to cold positive regulation of protein localization to nucleus negative regulation of translation in response to endoplasmic reticulum stress regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway positive regulation of glutathione biosynthetic process response to manganese-induced endoplasmic reticulum stress translational initiation response to oxidative stress proteasome-mediated ubiquitin-dependent protein catabolic process mitochondrial respiratory chain supercomplex assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch