Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
MA5-17161 targets RAB4A in FACS, IHC, indirect ELISA, and WB applications and shows reactivity with Human, Mouse, and Non-human primate samples.
The MA5-17161 immunogen is purified recombinant fragment of human RAB4A expressed in E. Coli.
MA5-17161 detects RAB4A which has a predicted molecular weight of approximately 23.8kDa.
Protein transport. Probably involved in vesicular traffic.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HTLV-1 related endogenous sequence; unnamed protein product
Gene Aliases: HRES-1; HRES-1/RAB4; HRES1; RAB4
UniProt ID: (Human) P20338, (Mouse) P56371
Entrez Gene ID: (Human) 5867, (Mouse) 19341
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support