Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Peptide Sequence: MEDKRSLSMA RCEERNSRGQ DHGLERVPFP PQLQSETYLH PADPSPAWDD PGSTGSPNLR LLTEEIAFQP LAEEASFRRP HPDGDVPPQG EDNLLSLPFP QKLWRLVSSN QFSSIWWDDS GACRVINQKL FEKEILKRDV AHKVFATTSI KSFFRQLNLY GFRKRRQCTF RTFTRIFSAK RLVSILNKLE FYCHPYFQRD SPHLLVRMKR RVGVKSAPRH QEEDKPEAAG SCLAPADTEQ QDHTSPNEND QVTPQHREPA GPNTQIRSGS APPATPVMVP DSAVASDNSP VTQPAGEWSE GSQAHVTPVA AVPGPAALPF LYVPGSPTQM NSYGPVVALP TASRSTLAMD TTGLPAPGML PFCHLWVPVT LVAAGAAQPA ASMVMFPHLP ALHHHCPHSH RTSQYMPASD GPQAYPDYAD QST
HSFX1 (Heat Shock Transcription Factor Family, X-Linked 1) is a Protein Coding gene. Diseases associated with HSFX1 include Microsporidiosis. Gene Ontology (GO) annotations related to this gene include DNA-binding transcription factor activity and sequence-specific DNA binding. An important paralog of this gene is HSFX2. [GeneCards]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ORC4L; testicular secretory protein Li 21
Gene Aliases: LW-1
UniProt ID: (Human) Q9UBD0
Entrez Gene ID: (Human) 100506164
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support